Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34574

Supelco

TISAB IV solution

for determination of fluoride

Synonym(s):

Total Ionic Strength Adjustment Buffer IV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

grade

for determination of fluoride

Quality Level

form

liquid

pH

5.6-6.0(20 °C)

General description

Total ionic strength adjustment buffers (TISAB), is generally used for the microdetermination of fluorides.
Visit our Sensor Applications portal to learn more.

Application

TISAB IV was used as a metal ion complexing and ionic concentration adjustment agent, in an experiment investigating the increase in fluoride release, from glass ionomer cement, after coating the discs of glass ionomer with 10% silver fluoride (AgF).

Other Notes

Important notice
The article number 34574-4X2.5L will be discontinued. Please order the single bottle 34574-2.5L which is physically identical with the same exact specifications.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microdetermination of fluoride using Gran's plots.
Microchimica Acta, 61 (1), 887-100 (1973)
Z Ariffin et al.
Australian dental journal, 51(4), 328-332 (2007-01-30)
This study investigated the extent to which a coating of 10% silver fluoride (AgF) on discs of glass jonomer cements (GIGs) would enhance the release of fluoride ion into eluting solutions at varying pH. Forty discs each of Fuji LX

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service