Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

34352

Supelco

Mephosfolan

PESTANAL®, analytical standard

Synonym(s):

Diethyl (4-methyl-1,3-dithiolan-2-ylidene)phosphoramidate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16NO3PS2
CAS Number:
Molecular Weight:
269.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)\N=C1/SCC(C)S1

InChI

1S/C8H16NO3PS2/c1-4-11-13(10,12-5-2)9-8-14-6-7(3)15-8/h7H,4-6H2,1-3H3/b9-8+

InChI key

LTQSAUHRSCMPLD-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Mephosfolan may be used as an analytical reference standard for the determination of the analyte in tea, grain, fruit- and vegetable-based infant foods and tobacco samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An effective method for pesticide residues determination in tobacco by GC-MS/MS and UHPLC-MS/MS employing acetonitrile extraction with low-temperature precipitation and d-SPE clean-up.
Bernardi G, et al.
Talanta, 161(1), 40-47 (2016)
Applications of LC/ESI-MS/MS and UHPLC/Qq-TOF-MS for the determination of 141 pesticides in tea
Wang J, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 94(6), 1685-1714 (2011)
N M Bakry et al.
Journal of analytical toxicology, 4(4), 212-214 (1980-07-01)
A sequential thin-layer chromatographic method (STLC) has been developed to analyze the two organophosphorus insecticides phosfolan (O,O-diethyl 1,3-dithiolan-2-ylidenephosphoramidate) and mephospholan (O,O-diethyl 4-methyl-1,3-dithiolan-2-ylidenephosphoramidate) and some of their possible degradation products. Gelman type SA, Instant Thin Layer Chromatography (ITLC) silicic acid-impregnated glass
Simultaneous determination of 405 pesticide residues in grain by accelerated solvent extraction then gas chromatography-mass spectrometry or liquid chromatography-tandem mass spectrometry.
Pang GF, et al.
Analytical and Bioanalytical Chemistry, 384(6), 1366-1408 (2006)
Determination of 142 pesticides in fruit-and vegetable-based infant foods by liquid chromatography/electrospray ionization-tandem mass spectrometry and estimation of measurement uncertainty.
Wang J and Leung D
Journal of AOAC (Association of Official Analytical Chemists) International, 92(1), 279-301 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service