Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34087

Supelco

Diethofencarb

PESTANAL®, analytical standard

Synonym(s):

Isopropyl N-(3,4-diethoxyphenyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21NO4
CAS Number:
Molecular Weight:
267.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOc1ccc(NC(=O)OC(C)C)cc1OCC

InChI

1S/C14H21NO4/c1-5-17-12-8-7-11(9-13(12)18-6-2)15-14(16)19-10(3)4/h7-10H,5-6H2,1-4H3,(H,15,16)

InChI key

LNJNFVJKDJYTEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethofencarb is a systemic, phenyl-carbamate fungicide, used for the control of gray mold on fruits and vegetables. Its mode of action involves the inhibition of phospholipids and fatty acid biosynthesis in pathogens.

Application

Diethofencarb may be used as an analytical reference standard for the determination of the analyte in peel and pulp of apples using dispersive liquid-liquid microextraction, followed by high-performance liquid chromatography with diode-array detection (HPLC-DAD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Cheng et al.
International journal of environmental analytical chemistry, 90(11), 845-855 (2010-09-24)
Dispersive liquid-liquid microextraction (DLLME) has been developed for the extraction and preconcentration of diethofencarb (DF) and pyrimethanil (PM) in environmental water. In the method, a suitable mixture of extraction solvent (50 μL carbon tetrachloride) and dispersive solvent (0.75 mL acetonitrile)
D W Hollomon et al.
Antimicrobial agents and chemotherapy, 42(9), 2171-2173 (1998-09-16)
Benzimidazoles are important antitubulin agents used in veterinary medicine and plant disease control. Resistance is a practical problem correlated with single amino acid changes in beta-tubulin and is often linked to greater sensitivity to phenylcarbamates. This negative cross-resistance creates opportunities
M Fujimura et al.
Current genetics, 21(4-5), 399-404 (1992-04-01)
Two MBC-resistant mutants of Neurospora crassa, F914 and F939, were sensitive to diethofencarb at a concentration of 0.1 micrograms/ml, while the wild-type strain and other MBC-resistant mutants showed resistance to diethofencarb at a concentration of 100 micrograms/ml. Genetic analysis suggested
K Saito et al.
Xenobiotica; the fate of foreign compounds in biological systems, 21(5), 575-582 (1991-05-01)
1. Age-related changes in metabolism of diethofencarb (DFC) (isopropyl-1,4-diethoxycarbanilate) and hepatic drug-metabolizing enzyme activities were studied in male Sprague-Dawley rats aged 2, 8, 16 and 21 months. 2. Following a single oral dose of 14C-DFC (500 mg/kg), the major route
S Hosokawa et al.
The Journal of toxicological sciences, 17(3), 155-166 (1992-08-01)
To examine the mechanism and toxicological significance of thyroidal tumor observed slightly in a long-term rat study with diethofencarb (isopropyl 3,4-diethoxycarbanilate), male Sprague-Dawley rats were fed diethofencarb in diets at concentrations of 0, 5,000 or 20,000 ppm for 3 months.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service