Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

34023

Supelco

2,4,6-Trichloroanisole-d5

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6D2OCD3
CAS Number:
Molecular Weight:
216.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

132 °C/28 mmHg (lit.)

mp

60-62 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

mass shift

M+5

SMILES string

[2H]c1c(Cl)c([2H])c(Cl)c(OC([2H])([2H])[2H])c1Cl

InChI

1S/C7H5Cl3O/c1-11-7-5(9)2-4(8)3-6(7)10/h2-3H,1H3/i1D3,2D,3D

InChI key

WCVOGSZTONGSQY-RHIBPKLGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,6-Trichloroanisole-d5 is a deuterium labeled analog of 2,4,6-trichloroanisole belonging to the class of chloroanisole, a major contaminant in wine industry.

Application

2,4,6-Trichloroanisole-d5 may be used as an internal standard for the quantification of 2,4,6-trichloroanisole in drinking water samples using gas chromatography coupled to mass spectrometry equipped with a thermal desorption unit (TDSA) and a PTV (programmable temperature vaporizer) injector.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On-tap passive enrichment, a new way to investigate off-flavor episodes in drinking water.
Tondelier C, et al.
Journal of Chromatography A, 1216(14), 2854-2859 (2009)
Navideh Sadoughi et al.
Journal of agricultural and food chemistry, 63(11), 2877-2885 (2015-02-24)
An optimized method for the quantitation of volatile compounds responsible for off-aromas, such as earthy odors, found in wine and grapes was developed. The method involved a fast and simple headspace solid-phase microextraction-gas chromatography-mass spectrometry (HS-SPME-GC-MS) for simultaneous determination of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service