Skip to Content
MilliporeSigma
All Photos(1)

Documents

32384

Supelco

Flumequine-(1,2,carboxy-13C3)

VETRANAL®, analytical standard

Synonym(s):

9-Fluoro-6,7-dihydro-5-methyl-1-oxo-1H,5H-benzo[ij]quinolizine-1,2-13C2-2-carboxylic acid-13C, Flumequine-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C11H12FNO3
CAS Number:
Molecular Weight:
264.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

FC1=CC2=C(N3C(C)CC2)C([13C]([13C]([13C](O)=O)=C3)=O)=C1

InChI

1S/C14H12FNO3/c1-7-2-3-8-4-9(15)5-10-12(8)16(7)6-11(13(10)17)14(18)19/h4-7H,2-3H2,1H3,(H,18,19)/i11+1,13+1,14+1

InChI key

DPSPPJIUMHPXMA-AIGZGMKWSA-N

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julieta Griboff et al.
Food chemistry, 332, 127380-127380 (2020-07-01)
The occurrence of 46 antibiotics (amphenicols, cephalosporins, dihydrofolate reductase inhibitors, fluroquinolones, macrolides, nitrofurans, penicillins, quinolones, sulfamides and tetracyclines) in Argentinean market fish were investigated by UPLC-MS/MS. Veterinary and human antimicrobials enrofloxacin, clarithromycin, roxithromycin, doxycycline and oxytetracycline were detected in 100%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service