Skip to Content
MilliporeSigma
All Photos(1)

Documents

31591

Supelco

Ampicillin trihydrate

VETRANAL®, analytical standard

Synonym(s):

D-(−)-α-Aminobenzylpenicillin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O4S · 3H2O
CAS Number:
Molecular Weight:
403.45
Beilstein:
5399534
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white to off-white

mp

198-200 °C (dec.) (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

clinical testing

format

neat

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C(O)=O

InChI

1S/C16H19N3O4S.3H2O/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;;;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);3*1H2/t9-,10-,11+,14-;;;/m1.../s1

InChI key

RXDALBZNGVATNY-CWLIKTDRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ampicillin trihydrate is a stable hydrated form of ampicillin.
Chemical structure: ß-lactam

Application

Ampicillin trihydrate has been used as reference standard for determining the residual solvents and its effect on ampicillin trihydrate crystal structure using GC, X-ray powder diffraction (XRPD) and Fourier transform infrared spectroscopy (FT-IR).
Commission Regulation (EU) No 37/2010 of 22 December 2009 on pharmacologically active substances and their classification regarding maximum residue limits in foodstuffs of animal origin
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

A β-lactam antibiotic with an amino group side chain attached to the penicillin structure. Penicillin derivative that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Bactericidal only to growing Escherichia coli . Mode of resistance: Cleavage of β-lactam ring of ampicillin by β-lactamase. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.

Packaging

250 mg

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of residual solvents and investigation of their effect on ampicillin trihydrate crystal structure.
Nojavan S
Journal of Pharmaceutical and Biomedical Analysis, 4;36(5), 983-988 (2005)
Ramendra K Singh et al.
European journal of medicinal chemistry, 45(3), 1078-1086 (2009-12-26)
Curcumin bioconjugates, viz. di-O-tryptophanylphenylalanine curcumin (2), di-O-decanoyl curcumin (3), di-O-pamitoyl curcumin (4), di-O-bis-(gamma,gamma)folyl curcumin (6), C(4)-ethyl-O-gamma-folyl curcumin (8) and 4-O-ethyl-O-gamma-folyl curcumin (10) have been synthesized and tested for their antibacterial and antiviral activities. The conjugates 2, 3, 4, 6 and
Sherif A F Rostom et al.
Bioorganic & medicinal chemistry, 17(2), 882-895 (2008-12-17)
The synthesis of two groups of structure hybrids comprising basically the antipyrine moiety attached to either polysubstituted thiazole or 2,5-disubstituted-1,3,4-thiadiazole counterparts through various linkages is described. Twelve out of the newly synthesized compounds were evaluated for their anti-inflammatory activity using
Ebtehal S Al-Abdullah et al.
European journal of medicinal chemistry, 46(9), 4642-4647 (2011-08-19)
New series of 6-phenyl-2,4-disubstituted pyrimidine-5-carbonitriles namely, 2-substitued thio-6-phenyl-3,4-dihydro-4-oxopyrimidine-5-carbonitriles (5a-d, 6, 7a-d, 8), 2-(4-chlorobenzylthio)-4-chloro-6-phenylpyrimidine-5-carbonitrile (9), 2-(4-chlorobenzylthio)-4-arylthio-6-phenylpyrimidine-5-carbonitriles (10a-d) and 2-(4-chlorobenzylthio)-4-arylamino-6-phenylpyrimidine-5-carbonitriles (11a-d) was synthesized and tested for in vitro activities against a panel of Gram-positive and Gram-negative bacteria and the yeast-like pathogenic fungus
Sherif A F Rostom et al.
Bioorganic & medicinal chemistry, 17(6), 2410-2422 (2009-03-03)
The azole pharmacophore is still considered a viable lead structure for the synthesis of more efficacious and broad spectrum antimicrobial agents. Potential antibacterial and antifungal activities are encountered with some tetrazoles. Therefore, this study presents the synthesis and antimicrobial evaluation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service