Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31237

Supelco

Dichloroprop-P

PESTANAL®, analytical standard

Synonym(s):

(R)-(+)-2-(2,4-Dichlorophenoxy)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8Cl2O3
CAS Number:
Molecular Weight:
235.06
Beilstein:
5265110
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C[C@@H](Oc1ccc(Cl)cc1Cl)C(O)=O

InChI

1S/C9H8Cl2O3/c1-5(9(12)13)14-8-3-2-6(10)4-7(8)11/h2-5H,1H3,(H,12,13)/t5-/m1/s1

InChI key

MZHCENGPTKEIGP-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloroprop-P is a selective, systemic and post-emergence herbicide, used for weed control management.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pesticide Encyclopedia (2014)
Yuezhong Wen et al.
Environmental science & technology, 44(13), 4981-4987 (2010-06-12)
Chitosan, a natural polysaccharide, is widely regarded as a biocompatible and potential carrier for controlled-release drug formulations. However, the potential effect of chitosan on the environmental behavior of contaminants is poorly understood, especially on chiral chemicals. In this study, the
Yuezhong Wen et al.
Environmental science & technology, 45(11), 4778-4784 (2011-05-07)
Reactive oxygen species (ROS) are considered to be the key players in cell toxicity. However, cross talk between the enantioselective toxicity of pesticides, heavy metals, and ROS is poorly understood. To decipher the puzzle, the effects of copper (Cu) on
Mélanie M Paulin et al.
Environmental microbiology, 13(6), 1513-1523 (2011-03-23)
We investigated the effect of (R,S)-dichlorprop herbicide addition to soil microcosms on the degrading indigenous microbial community by targeting multiple α-ketoglutarate-dependent (α-KG) dioxygenase-encoding genes (rdpA, sdpA and tfdA group I) at both gene and transcript level. The soil microbial community
Yuezhong Wen et al.
Journal of environmental monitoring : JEM, 13(4), 879-885 (2011-02-08)
To reduce the leaching potential, to prevent groundwater contamination and to maintain the efficacy of a pesticide, natural polysaccharides have received increasing attention due to their biocompatibility and useful biological reactivity for controlled release formulations (CRFs) of pesticides. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service