Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

21856

Supelco

Carbonate ionophore IV solution

in THF (50 mg/0.5 mL), Selectophore

Synonym(s):

4-Butyl-α,α,α-trifluoroacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13F3O
CAS Number:
Molecular Weight:
230.23
Beilstein:
2105281
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

concentration

in THF (50 mg/0.5 mL)

SMILES string

CCCCc1ccc(cc1)C(=O)C(F)(F)F

InChI

1S/C12H13F3O/c1-2-3-4-9-5-7-10(8-6-9)11(16)12(13,14)15/h5-8H,2-4H2,1H3

InChI key

XIVOMWSWGCFRNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Carbonate ionophore IV solution (4-Butyl-a,a,a-trifluoroacetophenone) was used in optimization of liquid membrane-cellophane electrodes to develop a carbonate-selective electrode for direct measurement of CO2 at physiological pH.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Physical form

solution in 0.5 mL tetrahydrofuran

Other Notes

Lipophilic, neutral ionophore for the determination of carbonate and hydroxide with solvent polymeric membrane electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johan Bobacka et al.
Biosensors & bioelectronics, 18(2-3), 245-253 (2002-12-18)
A carbonate ion-selective electrode for determination of total carbon dioxide species such as carbon dioxide, bicarbonate and carbonate with reduced interference from salicylate is described. Derivatives of trifluoroacetophenone were used as neutral carriers for carbonate. A polymer-free liquid carbonate-selective membrane
Role of trifluoroacetophenone solvents and quaternary ammonium salts in carbonate-selective liquid membrane electrodes
Meyerhoff, M.E., et al.
Analytical Chemistry, 59 (1), 144-150 (1987)
O Dinten et al.
Analytical chemistry, 63(6), 596-603 (1991-03-15)
On the basis of previously reported correlations between the lipophilicity of membrane components, their partition coefficient between the membrane and the sample, and the lifetime of corresponding neutral-carrier-based sensors, the lipophilicities of ionophores and plasticizers in analytically relevant ion-selective electrodes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service