Skip to Content
MilliporeSigma
All Photos(2)

Documents

201960

Sigma-Aldrich

Acid Yellow 25

Dye content 40 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N5NaO6S2
CAS Number:
Molecular Weight:
549.55
Colour Index Number:
18835
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 40%

λmax

392 nm

ε (extinction coefficient)

13200 at 259 nm in H2O at 0.03 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1ccc(cc1S(=O)(=O)Nc2ccccc2)\N=N\c3c(C)nn(c3O)-c4ccc(cc4)S([O-])(=O)=O

InChI

1S/C23H21N5O6S2.Na/c1-15-8-9-18(14-21(15)35(30,31)27-17-6-4-3-5-7-17)24-25-22-16(2)26-28(23(22)29)19-10-12-20(13-11-19)36(32,33)34;/h3-14,27,29H,1-2H3,(H,32,33,34);/q;+1/p-1/b25-24+;

InChI key

VKLQVFKYJGXGKQ-QREUMGABSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shumin Xia et al.
Cell reports, 28(5), 1251-1267 (2019-08-01)
Mechanical cues influence pluripotent stem cell differentiation, but the underlying mechanisms are not well understood. Mouse embryonic stem cells (mESCs) exhibit unusual cytomechanical properties, including low cell stiffness and attenuated responses to substrate rigidity, but the underlying structural basis remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service