Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

112763

Sigma-Aldrich

Diphenylamine

ReagentPlus®, 99%

Synonym(s):

N-Phenylbenzenamine, DBA, DFA, DPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NH
CAS Number:
Molecular Weight:
169.22
Beilstein:
508755
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

5.82 (vs air)

Quality Level

vapor pressure

1 mmHg ( 108 °C)

product line

ReagentPlus®

Assay

99%

autoignition temp.

1175 °F

bp

302 °C (lit.)

mp

50-53 °C (lit.)

solubility

water: insoluble

SMILES string

N(c1ccccc1)c2ccccc2

InChI

1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H

InChI key

DMBHHRLKUKUOEG-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

General description

Diphenylamine, a secondary aromatic amine, is an N-substituted derivative of aniline.

Application

Diphenylamine may be used in the preparation of diphenylamine reagent, which is employed for the quantitative estimation of nucleic acid (DNA) from various biological sources by colorimetric method. It may be used in the preparation of poly(diphenylamine), via electrochemical and chemical polymerization methods. It can also be used as an internal standard in the determination of hair nicotine via gas chromatography coupled with mass spectrometry.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney,Liver,spleen

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Determination of hair nicotine by gas chromatography?mass spectrometry"
Man NC, et al
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 877(03), 339-342 (2009)
An improved diphenylamine method for the estimation of deoxyribonucleic acid.
Giles KW and Myers A.
Nature, 206(4979) , 93-93 (1965)
A study of the conditions and mechanism of the diphenylamine reaction for the colorimetric estimation of deoxyribonucleic acid.
K BURTON
The Biochemical journal, 62(2), 315-323 (1956-02-01)
Soluble and methane sulfonic acid doped poly (diphenylamine)-synthesis and characterization.
Wen T-C, et al.
Materials Letters, 57(2), 280-290 (2002)
Walter J Jessen et al.
The Journal of clinical investigation, 123(1), 340-347 (2012-12-12)
Neurofibromatosis type 1 (NF1) patients develop benign neurofibromas and malignant peripheral nerve sheath tumors (MPNST). These incurable peripheral nerve tumors result from loss of NF1 tumor suppressor gene function, causing hyperactive Ras signaling. Activated Ras controls numerous downstream effectors, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service