Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 mL | Check Cart for Availability | $101.00 |
About This Item
$101.00
Quality Segment
product line
TraceCERT®
grade
certified reference material, TraceCERT®
concentration
1000 mg/L butyrate in water (nominal concentration)
technique(s)
ion chromatography: suitable
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
format
single component solution
General description
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Analysis Note
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable |
| application(s) cleaning products | application(s) environmental | application(s) environmental | application(s) environmental |
| grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® |
| format single component solution | format single component solution | format single component solution | format single component solution |
| concentration 1000 mg/L butyrate in water (nominal concentration) | concentration 1000 mg/L bromate in water (nominal concentration) | concentration 1000 mg/L in water (nominal concentration) | concentration 1000 mg/L in water (nominal concentration) |
| product line TraceCERT® | product line TraceCERT® | product line TraceCERT® | product line TraceCERT® |
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



