Skip to Content
MilliporeSigma
All Photos(1)

Documents

06234

Sigma-Aldrich

Aluminum phosphate monobasic solution

~50% in H2O

Synonym(s):

Aluminum dihydrogen phosphate, prim-Aluminum phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(H2PO4)3
CAS Number:
Molecular Weight:
317.94
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

concentration

~50% in H2O

Quality Level

density

1.5 g/mL at 20 °C

SMILES string

[Al+3].OP(O)([O-])=O.OP(O)([O-])=O.OP(O)([O-])=O

InChI

1S/Al.3H3O4P/c;3*1-5(2,3)4/h;3*(H3,1,2,3,4)/q+3;;;/p-3

InChI key

RGPUVZXXZFNFBF-UHFFFAOYSA-K

Application

Aluminum phosphate monobasic solution can be used to prepare a Ni-based catalyst named Ni-SAPO-34, which is used in the conversion of methanol to olefins.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of a catalytically active nickel silicoaluminophosphate catalyst for the conversion of methanol to ethene
Thomas JM, et al.
Chemistry of Materials, 3(4), 667-672 (1991)
Belma Ljutic et al.
Vaccine, 30(19), 2981-2988 (2012-03-03)
We investigated the immunogenicity, stability and adsorption properties of an experimental pneumococcal vaccine composed of three protein vaccine antigens; Pneumococcal histidine triad protein D, (PhtD), Pneumococcal choline-binding protein A (PcpA) and genetically detoxified pneumolysin D1 (PlyD1) formulated with aluminum salt
Shibo Jiang et al.
Expert review of vaccines, 11(12), 1405-1413 (2012-12-21)
A subunit vaccine, RBD-S, is under development to prevent severe acute respiratory syndrome (SARS) caused by SARS coronavirus (SARS-CoV), which is classified by the US NIH as a category C pathogen. This vaccine is comprised of a recombinant receptor-binding domain
Ruth Pepper et al.
BMC nephrology, 12, 55-55 (2011-10-14)
Aluminium (Al) toxicity was frequent in the 1980s in patients ingesting Al containing phosphate binders (Alucaps) whilst having HD using water potentially contaminated with Al. The aim of this study was to determine the risk of Al toxicity in HD
Wenlei Xie et al.
Bioresource technology, 119, 60-65 (2012-06-26)
WO(3)/AlPO(4) catalysts were prepared by impregnation of AlPO(4) with ammonium metatungstate. Powder X-ray diffraction (XRD), Fourier transform infrared (FT-IR) spectroscopy, and thermo gravimetric and differential thermal analysis (TG-DTA) demonstrated that the tungsten compound was incorporated into AlPO(4) forming the catalyst

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service