Skip to Content
MilliporeSigma
All Photos(1)

Documents

04314

Supelco

Thujone Standard Mixture

analytical standard

Synonym(s):

α,β-Thujone, (−)-1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

description

reference substance for GC

composition

α-thujone, 90-98%
β-thujone, 2-10%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.450
n20/D 1.455 (lit.)

bp

84-86 °C/17 mmHg (lit.)

density

0.925 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C)[C@@]12C[C@@H]1[C@@H](C)C(=O)C2.CC(C)[C@@]34C[C@@H]3[C@H](C)C(=O)C4

InChI

1S/2C10H16O/c2*1-6(2)10-4-8(10)7(3)9(11)5-10/h2*6-8H,4-5H2,1-3H3/t7-,8+,10-;7-,8-,10+/m01/s1

InChI key

AKPBIVZAUPVDMO-APEKEWONSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This compound is commonly found in plants of the genus: achillea salvia

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service