Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-5290

Sigma-Aldrich

Potassium phosphate monobasic

≥99.5%

Synonym(s):

Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

≥99.5%

form

crystalline

availability

available only in Japan

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

252.6 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Suitability

pH standard solution

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Przemyslaw Nogly et al.
IUCrJ, 2(Pt 2), 168-176 (2015-04-14)
Lipidic cubic phases (LCPs) have emerged as successful matrixes for the crystallization of membrane proteins. Moreover, the viscous LCP also provides a highly effective delivery medium for serial femtosecond crystallography (SFX) at X-ray free-electron lasers (XFELs). Here, the adaptation of
Pamela Gamba et al.
PLoS genetics, 11(6), e1005047-e1005047 (2015-06-26)
Gene expression can be highly heterogeneous in isogenic cell populations. An extreme type of heterogeneity is the so-called bistable or bimodal expression, whereby a cell can differentiate into two alternative expression states. Stochastic fluctuations of protein levels, also referred to
Suvi Vartiainen et al.
Disease models & mechanisms, 7(6), 635-648 (2014-05-09)
A point mutation in the Drosophila gene that codes for the major adult isoform of adenine nuclear translocase (ANT) represents a model for human diseases that are associated with ANT insufficiency [stress-sensitive B(1) (sesB(1))]. We characterized the organismal, bioenergetic and
Megan A Rees et al.
Journal of proteome research, 14(1), 120-132 (2014-10-21)
Interactions between a host and a bacterial pathogen are mediated by cross-talk between molecules present on, or secreted by, pathogens and host binding-molecules. Identifying proteins involved at this interface would provide substantial insights into this interaction. Although numerous studies have
Kirsi Penttinen et al.
PloS one, 10(5), e0125366-e0125366 (2015-05-09)
Catecholaminergic polymorphic ventricular tachycardia (CPVT) is a highly malignant inherited arrhythmogenic disorder. Type 1 CPVT (CPVT1) is caused by cardiac ryanodine receptor (RyR2) gene mutations resulting in abnormal calcium release from sarcoplasmic reticulum. Dantrolene, an inhibitor of sarcoplasmic Ca(2+) release

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service