Skip to Content
MilliporeSigma
All Photos(1)

Documents

05-4535

Sigma-Aldrich

Chromium(III) oxide

SAJ first grade, ≥99.0%

Synonym(s):

Chromia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr2O3
CAS Number:
Molecular Weight:
151.99
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.0%

form

powder

availability

available only in Japan

SMILES string

O=[Cr]O[Cr]=O

InChI

1S/2Cr.3O

InChI key

QDOXWKRWXJOMAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in a solid-state reaction to form the new ternary oxide, CrVMoO7, can be used in selective oxidation.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poornima Vajpayee et al.
Journal of biomedical nanotechnology, 7(1), 205-206 (2011-04-14)
In this study, seeds of Triticum aestivum L. (Poaceae) were exposed to 0-100 microg/mL chromium oxide nanoparticles (Cr2O3, Nps) to study the phytotoxic effects on seed germination and seedling growth. It has been observed that 25-100 microg/mL Cr2O3, Nps inhibited
Noriko Nishino et al.
Physical chemistry chemical physics : PCCP, 14(45), 15840-15848 (2012-10-24)
While many studies of heterogeneous chemistry on Cr(2)O(3) surfaces have focused on its catalytic activity, less is known about chemistry on this surface under atmospheric conditions. We report here studies of the thermal and photochemical reactions of NO(2) on Cr(2)O(3)
Nien-Hsun Li et al.
Journal of hazardous materials, 198, 356-361 (2011-11-15)
The feasibility of stabilizing nickel-laden sludge by a high-temperature NiCr(2)O(4) synthesis process was investigated with different sintering temperatures, salt contents, molar ratios, and reaction atmospheres. The crystalline phases of species were investigated by using an X-ray diffraction, and the surface
Isabella Marr et al.
Sensors (Basel, Switzerland), 11(8), 7736-7748 (2011-12-14)
Zeolites are promising materials in the field of gas sensors. In this technology-oriented paper, a planar setup for potentiometric hydrocarbon and hydrogen gas sensors using zeolites as ionic sodium conductors is presented, in which the Pt-loaded Na-ZSM-5 zeolite is applied
Selvam Sangeetha et al.
Colloids and surfaces. B, Biointerfaces, 100, 36-41 (2012-07-07)
Stabilization of collagen for various applications employs chemicals such as aldehydes, metal ions, polyphenols, etc. Stability against enzymatic, thermal and mechanical degradation is required for a range of biomedical applications. The premise of this research is to explore the use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service