Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03-1730

Sigma-Aldrich

Benzyl benzoate

SAJ first grade, ≥98.0%

Synonym(s):

Benzoic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH2C6H5
CAS Number:
Molecular Weight:
212.24
Beilstein:
2049280
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

1 mmHg ( 125 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

896 °F

availability

available only in Japan

refractive index

n20/D 1.568 (lit.)

bp

323-324 °C (lit.)

mp

17-20 °C (lit.)

density

1.118 g/mL at 20 °C (lit.)

SMILES string

O=C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

SESFRYSPDFLNCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

298.4 °F

Flash Point(C)

148 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osamu Ohno et al.
Bioorganic & medicinal chemistry, 16(16), 7843-7852 (2008-08-02)
Hypertension is a lifestyle-related disease which often leads to serious conditions such as heart disease and cerebral hemorrhage. Angiotensin II (Ang II) plays an important role in regulating cardiovascular homeostasis. Consequently, antagonists that block the interaction of Ang II with
Ben Spitzer-Rimon et al.
The Plant cell, 22(6), 1961-1976 (2010-06-15)
Floral scent, which is determined by a complex mixture of low molecular weight volatile molecules, plays a major role in the plant's life cycle. Phenylpropanoid volatiles are the main determinants of floral scent in petunia (Petunia hybrida). A screen using
A K Charles et al.
Journal of applied toxicology : JAT, 29(5), 422-434 (2009-04-02)
Benzyl salicylate, benzyl benzoate and butylphenylmethylpropional (Lilial) are added to bodycare cosmetics used around the human breast. We report here that all three compounds possess oestrogenic activity in assays using the oestrogen-responsive MCF7 human breast cancer cell line. At 3
Mayumi Bowen et al.
Journal of pharmaceutical sciences, 101(12), 4433-4443 (2012-09-25)
Developing high-concentration monoclonal antibody (mAb) liquid formulations for subcutaneous (s.c.) administration is challenging because increased viscosity makes injection difficult. To overcome this obstacle, we investigated a nonaqueous powder suspension approach. Three IgG1 mAbs were spray dried and suspended at different
Catherine W McCollum et al.
Aquatic toxicology (Amsterdam, Netherlands), 152, 152-163 (2014-04-29)
Exposure to arsenic in its inorganic form, arsenite, causes adverse effects to many different organs and tissues. Here, we have investigated arsenite-induced adverse effects on vascular tissues in the model organism zebrafish, Danio rerio. Zebrafish embryos were exposed to arsenite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service