Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01-4960

Sigma-Aldrich

Ammonium phosphate monobasic

JIS special grade, ≥99.0%

Synonym(s):

mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

vapor pressure

<0.000 hPa ( 20 °C)

Assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

3.8-4.4 (25 °C, 50 g/L)

mp

190 °C (dec.) (lit.)

SMILES string

N.OP(O)(O)=O

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Young Yang et al.
Clinical chemistry and laboratory medicine, 53(8), 1249-1257 (2015-02-27)
Since the urinary concentration of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol (NNAL) is a reliable biomarker of exposure to tobacco smoke, we developed a relatively simple high-throughput chromatographic method to quantify total urinary NNAL concentrations in the general population. The high-throughput analytical method was developed
Maria Lina Sanna et al.
Fungal biology, 116(7), 769-777 (2012-07-04)
Pichia fermentans DiSAABA 726 is a biofilm-forming yeast that undergoes dimorphic transition. Under yeast-like morphology it controls brown rot caused by Monilia spp. on apple fruit, while under pseudohyphal form, it shows pathogenic behaviour itself on peach fruit. The present
Everest Akaraonye et al.
Biotechnology journal, 7(2), 293-303 (2011-12-08)
The main hindrance in the use of polyhydroxyalkanoates (PHAs) as a replacement for existing petroleum-based plastics is their high production cost. The carbon source accounts for 50% of the cost for PHA production. Thus, increasing the yield and productivity of
Xiaopeng Gao et al.
Journal of the science of food and agriculture, 90(5), 750-758 (2010-04-01)
A 3-year field trial was conducted to investigate the effect of agricultural management practices including tillage, preceding crop and phosphate fertilization on root colonization by arbuscular mycorrhizal (AM) fungi and grain cadmium (Cd) concentration of durum wheat (Triticum turgidum L.).
G A Morán-Marroquín et al.
International journal of food microbiology, 151(1), 87-92 (2011-09-10)
Knowledge of physiological behavior of indigenous tequila yeast used in fermentation process is still limited. Yeasts have significant impact on the productivity fermentation process as well as the sensorial characteristics of the alcoholic beverage. For these reasons a better knowledge

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service