Key Documents
SYMBISL01WW
Millipore
Steritest Symbio ISL Pump Kit, 2 media
About This Item
UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
JA.12
material
316L stainless steel
Quality Level
product line
Steritest®
weight
17.6 kg (pump body) with standard accessories:19.2 kg
General description
The Steritest Symbio ISL Pump is optimized for more ergonomic sterility testing inside isolators. Its table-integrated design offers more working space and loading volume in isolators.
Application
The Steritest Symbio ISL Pump, designed to fit your testing environment, Sterility Testing.
Legal Information
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Sterility testing USP 71, EP2.6.1 , JP 4.06