Key Documents
STC11FHA
Millipore
1/4 in. Lynx® ST Connector
About This Item
UNSPSC Code:
41121815
eCl@ss:
32010310
material
Lynx® polyetherimide connections (USP Class VI, 21CFR)
silicone seal (USP Class VI, 21CFR)
Quality Level
sterility
non-sterile
manufacturer/tradename
Lynx®
parameter
1.4 bar max. differential pressure (20 psid) at 4-40 °C (during actuation)
2 L/min flow rate at 0.1 bar
2.4 bar max. differential pressure (35 psid) at 4-40 °C
4 L/min flow rate at 0.4 bar
4-40 °C temperature
7 L/min flow rate at 1.3 bar
envelope L × W × H
81.3 mm (3.2 in.) × 40.6 mm (1.6 in.) × 68.6 mm (2.7 in.)
fitting
3/4 in. Tri-Clover inlet
1/4 in. outlet hose barb
Preparation Note
Sterilization Method
Gamma compatible up to 45 kGy; Steam @ 135 °C for 30 min (closed) 2 times
Legal Information
LYNX is a registered trademark of Merck KGaA, Darmstadt, Germany
Visit the Document Library
Articles
This article explains how to streamline transfer of an allogeneic cell therapies process to a CDMO partner and long-term manufacturing success.
This article explains how to streamline transfer of an allogeneic cell therapies process to a CDMO partner and long-term manufacturing success.
This article explains how to streamline transfer of an allogeneic cell therapies process to a CDMO partner and long-term manufacturing success.
This article explains how to streamline transfer of an allogeneic cell therapies process to a CDMO partner and long-term manufacturing success.