Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

STBMCTM12

Millipore

Clear Fluid Thioglycollate Medium

ready-to-use, bottle volume 100 mL , filling Volume

Synonym(s):

CTM, Clear FTM, Clear Thioglycollate Broth, Fluid Thioglycollate Medium (Clear), Thioglyc. Med. (FTM) clear, Thioglycollate B. 0.1l clear

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
eCl@ss:
32014001
NACRES:
NA.74

Agency

in accordance with EP 2.6.1
in accordance with JP 4.06
in accordance with USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

bottle of

bottle capacity

125 mL

bottle volume

100 mL , filling Volume

input

sample type pharmaceutical(s)

pH

7.1

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest®

shipped in

ambient

storage temp.

2-25°C

suitability

aerobic bacteria
nonselective for

General description

Clear Thioglycollate medium has the same growth promotion properties as the standard Fluid Thioglycollate Medium (FTM). The alternative formulation brings extra visual clarity versus the FTM which has a slight turbidity or haze. Composition and growth performance of this medium comply with the European Pharmacopeia (EP) 2.6.1, United States Pharmacopeia (USP) 71 and Japanese Pharmacopeia (JP) 4.06. Our manufacturing approach ensures the highest level of quality for our media, improving accuracy and significantly reducing the risk of incorrect interpretation and false results. The appearance of the fluid is light yellow and viscous liquid with a pink ring in suspension ≤1 cm. The pH of the product is 7.1 ± 0.2 and can be stored at 2-25°C for 12 months. The incubation temperature varies between 30-35°C, with an incubation time of 7-14 days. One box of product contains 12 bottles of 100mL of Medium. Each bottle has a screw cap with a septum that minimizes the risks of cross-contamination and optimizes the disinfection procedures.

Application

Clear Fluid Thioglycollate Medium is intended for the detection of anaerobic bacteria however, it also enables aerobic bacterial detection. This medium is used for sterility testing by membrane filtration or direct inoculation. The QC organisms include Clostridium sporogenes (ATCC 19404), Staphylococcus aureus (ATCC 6538) and Pseudomonas aeruginosa (ATCC 9027).
Sterility testing (membrane filtration and direct inoculation)
Bioburden testing

Components

Screw cap with septum

Specifications

Incubation Time: 7–14 days

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service