Skip to Content
MilliporeSigma
All Photos(1)

Documents

SPR00SIA2

Milli-Q

SmartPak® Purification Pack

For RiOs-DI® Systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

Quality Level

feature

A unique purification pak to produce DI water directly from tap water

packaging

pkg of 1 unit

manufacturer/tradename

SmartPak®

resistivity

>18.0 MΩ-cm (at 25 °C)

compatibility

for use with RiOs-DI® UV
for use with RiOs-DI®
for use with RiOs-DI® Clinical

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The SmartPak® RODI Purification Pack combines the purifications steps of reverse osmosis and ion exchange to produce type 2 pure water.
For use with RiOs-DI® water purification systems.

Application

This pack removes a wide variety of contaminants (organics, ions, particles...).

Features and Benefits

  • Genuine consumable for the replacement of SPR00SIA2 - SmartPak® Purification Pack.
  • Reliable and affordable type 2 pure water solution. 
  • Care-free maintenance: easily installed and replaced.
  • Certificate of Quality available online.

Legal Information

RIOS-DI is a registered trademark of Merck KGaA, Darmstadt, Germany
SMARTPAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service