Skip to Content
MilliporeSigma
All Photos(6)

Documents

SCR078

Sigma-Aldrich

Fluorescent Human ES/iPS Cell Characterization Kit

This Fluorescent Human ES/iPS Cell Characterization Kit contains a range of sensitive tools for the phenotypic assessment of the pluripotent status of human Embryonic stem & induced pluripotent Stem cells.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

technique(s)

immunocytochemistry: suitable

shipped in

wet ice

General description

Human embryonic stem (hES) cells are pluripotent cells derived from the inner cell mass of pre-implantation blastocysts. Human induced pluripotent (hiPS) cells are pluripotent cells generated by reprogramming human somatic cells using four transcription factors, Oct-4, Klf-4, Sox-2, and c-Myc, or their variants. Both hESC and hiPSC can self-renew and have the ability to generate all three germ layers: ectoderm, mesoderm, and endoderm. In vitro, hESC/iPSC are normally maintained and propagated on mouse fibroblast feeders for extended periods in media containing basic fibroblast growth factor (bFGF). However, spontaneous differentiation may occur in subpopulations of cells. Several pluripotent markers are commonly used to distinguish pluripotent hESC/iPSC from differentiated cells.

EMD Millipore’s Fluorescent Human ES/iPS Cell Characterization Kit contains a range of sensitive tools for the phenotypic assessment of the pluripotent status of human ES/iPS cells. Included in the kit is an enzymatic assay to measure alkaline phosphatase activity in the cells along with validated directly conjugated antibodies to pluripotent transcription factors, Oct-4, Sox-2 and Nanog and cell surface epitopes TRA-1-60 and TRA-1-81 to enable rapid immunocytochemical marker analysis. The Dapi nuclear dye is conveniently included to aid in cell quantification. While the expression levels of pluripotent markers are expected to be diminished upon differentiation, each possess specific expression kinetics. For example, it has been noted that upon differentiation, Oct-4 and TRA-1-60 expressions are the first to be down-regulated while Nanog and alkaline phosphatase down-regulate at a much slower timeframe.

Specificity

Cross Reactivty
None

Application

Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This Fluorescent Human ES/iPS Cell Characterization Kit contains a range of sensitive tools for the phenotypic assessment of the pluripotent status of human Embryonic stem & induced pluripotent Stem cells.

Components

1. Fast Red Violet solution (Part No. 90239): One 15 mL bottle.

2. Napthol AS-BI phosphate solution (Part No. 90234). One 15 mL bottle.

3. Mouse anti-Oct-4 (POU5f1), clone 7F9.2, Alexa Fluor 488 conjugate (Part No. MAB4419A4-50UL). One vial containing 50 µL of 0.5 mg/mL conjugated monoclonal antibody.

4. Mouse anti-Sox-2, clone 10H9.1, Cy3 conjugate (Part No. MAB4423C3-50UL). One vial containing 50 µL of 0.5 mg/mL conjugated monoclonal antibody.

5. Mouse anti-Nanog, clone 7F7.1, Alexa Fluor 488 conjugate (Part No. MABD24A4-50UL). One vial containing 50 µL of 0.5 mg/mL conjugated monoclonal antibody.

6. Mouse anti-TRA-1-60, clone TRA-1-60, Cy3 conjugate (Part No. MAB4360C3-50UL). One vial containing 50 µL of 0.5 mg/mL conjugated monoclonal antibody.

7. Mouse anti-TRA-1-81, clone TRA-1-81, Cy3 conjugate (Part No. MAB4381C3-50UL). One vial containing 50 µL of 0.5 mg/mL conjugated monoclonal antibody.

8. DAPI, 100 µL (Part No. 90229). One vial containing 100 µL volume.

Quality

Conjugated antibodies and alkaline phosphatase reagents have been validated on H9 human ES and STEMCCA derived human ips cells.

Physical form

Purified conjugated mouse monoclonal antibodies are stored in PBS with 0.1% sodium azide and 15mg/ml BSA.

Storage and Stability

Store at 2-8°C until expiration date of kit. Do not freeze or expose to elevated temperatures. Discard any remaining reagents after the expiration date.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa Conti Mazza et al.
Stem cell research, 55, 102506-102506 (2021-08-23)
Mutations in the oncogene PARK7, which codes for DJ-1, have been associated with early-onset autosomal recessive Parkinson's disease (PD); however, the exact role of DJ-1 in PD remains elusive. Fibroblasts from a PD patient with a uniparental disomy, 1 bp deletion

Protocols

Step-by-step stem cell culture protocols for human induced pluripotent stem cells (iPSCs) including ips cell thawing, expanding, freezing and characterizing.

Step-by-step stem cell culture protocols for human induced pluripotent stem cells (iPSCs) including ips cell thawing, expanding, freezing and characterizing.

Step-by-step stem cell culture protocols for human induced pluripotent stem cells (iPSCs) including ips cell thawing, expanding, freezing and characterizing.

Step-by-step stem cell culture protocols for human induced pluripotent stem cells (iPSCs) including ips cell thawing, expanding, freezing and characterizing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service