Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCM087

Sigma-Aldrich

Mouse iPS Reprogramming Boost Supplement

The Mouse iPS Reprogramming Boost Supplement enhances the efficiency of mouse iPS colony formation by at least 3-fold.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

Quality Level

form

liquid

input

sample type induced pluripotent stem cell(s)

shipped in

dry ice

General description

Small molecules targeting specific signaling pathways have been shown to enhance the efficiency and/or replace transcription factors involved in somatic cell reprogramming. EMD Millipore has screened multiple chemical compounds and selected the compounds in this kit based on their effects on enhancement of reprogramming efficiency as assessed by the number and overall quality of mouse iPS colonies generated.

The Mouse iPS Reprogramming Boost Supplement contains three proprietary small molecules in amounts sufficient to supplement over 250 mL mouse ES/iPS cell maintenance medium. When used in conjunction with the Mouse STEMCCA lentivirus reprogramming kits (SCR530, SCR531), the Mouse iPS Reprogramming Boost Supplement enhanced the efficiency of mouse iPS colony formation by at least 3-fold. Mouse iPS Reprogramming Boost Supplement can be used to enhance reprogramming efficiency in somatic cell lines that may be more refractory to reprogramming. Mouse iPS cells generated through use of the Mouse iPS Reprogramming Boost Supplement display characteristic ES cell-like morphology, stained positive for alkaline phosphatase, expressed the correct mouse ES cell marker phenotype (SSEA-1 and Sox-2) and can be rapidly expanded in normal mouse ES cell culture conditions.

Components

Kit contains sufficient quantities to supplement over 250 mL of medium.

1. TGF-ß RI Kinase Inhibitor II Supplement (1000X): (Part No. CS204458) One (1) vial containing 300 µL of the inhibitor in high quality DMSO.

2. GSK3ß Inhibitor Supplement (1000X): (Part No. CS204418) One (1) vial containing 300 µL of the inhibitor in high quality DMSO.

3. Methylation Modulator-A Supplement (1000X): (Part No. CS204419) One (1) vial containing 300 µL of the inhibitor in water.

Storage and Stability

Components of the Mouse iPS Reprogramming Boost Supplement are stable for at least 6 months from date of receipt when stored at -20°C. Upon first thaw, centrifuge the vial and gently mix the solution. Aliquot into smaller working volumes and freeze at -20°C or -80°C. Upon addition of the components of the Mouse iPS Cell Boost Supplement to the media, filter the supplemented media with a 0.22 µM filtration unit. Supplemented media is good for up to two weeks when stored at 2-8°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service