Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PCYTMAG23PMX13BK

Millipore

MILLIPLEX® Porcine Cytokine/Chemokine Magnetic Bead Panel Space Saver (Bulk) Packaging

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology in porcine serum, plasma and cell culture samples.

Synonym(s):

luminex porcine cytokine chemokine growth factor multiplex assay, luminex porcine pro inflammatory anti inflammatory cytokine panel, millipore porcine inflammation multiplex kit, porcine interleukin immunoassay panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

pig

manufacturer/tradename

Milliplex®

assay range

standard curve range: 0.005-20 ng/mL
(IL-1α)

standard curve range: 0.012-50 ng/mL
(IL-8)

standard curve range: 0.024-100 ng/mL
(GM-CSF, IL-1ra, IL-2, IL-6, IL-10, IL-12, IL-18 & TNFα)

standard curve range: 0.061-250 ng/mL
(IL-4)

standard curve range: 0.122-500 ng/mL
(IFNγ & IL-1β)

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

With a similarity to humans in heart size, blood flow rate, skin, liver enzymes and bone healing, porcine models have replaced many canine and non-human primate models in many research areas such as cardiovascular, toxicology, pharmacology, aging and bone. “Minipigs,” bred to retain the similar heart size and function as humans, but age without growing too large, have been used for long term preclinical or multi-generational studies which may be performed in a comparatively short period of time.

MILLIPLEX® Porcine Cytokine/Chemokine Panel can be used for the simultaneous quantification of all the following analytes in porcine tissue/cell lysate and culture supernatant samples and serum or plasma samples: GM-CSF, IFNγ, IL-1α, IL-1ra, IL-1β, IL-2, IL-4, IL-6, IL-8, IL-10, IL-12, IL-18 and TNFα. This kit uses a 96-well format, contains a lyophilized standard cocktail, two internal assay quality controls and can measure up to 38 samples in duplicate.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

UPDATE >>
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: GM-CSF, IFNγ, IL-1α, IL-1β, IL-1Ra, IL-2, IL-4, IL-6, IL-8 (CXCL8), IL-10, IL-12, IL-18, TNF-α
  • Recommended Sample Type: serum, plasma or cell/tissue culture supernatants or lysate
  • Recommended Sample Dilution: 25 μL per well of undiluted serum or plasma; cell/tissue culture samples may require dilution in an appropriate control medium.
  • NOTE: Extra centrifugation and attention may be needed for samples with extremely high lipid content.
  • Assay Run Time: Overnight (16-18 hours) at 2-8°C.
  • Research Category: Inflammation & Immunology

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service