Skip to Content
MilliporeSigma

P3C003C01

Millipore

Pellicon® 3 Cassette with Ultracel® Membrane

3 kDa, C screen, 0.11 m2

Synonym(s):

Pellicon® 3 Cassette with ULTRACEL® 3 kDa Membrane, C screen, 0.11 m2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602

material

composite regenerated cellulose (CRC) membrane
polypropylene (Screen (feed))
polypropylene cap (end cap)
polypropylene housing

Quality Level

product line

Pellicon®

storage condition

do not freeze

parameter

4-8 L/min-m2 flow rate

technique(s)

tangential flow filtration (TFF): suitable
ultrafiltration: suitable

H

2.4 cm (0.93 in.)

L

20.6 cm (8.1 in.)

filtration area

0.11 m2

size

68 mL , Volume (permeate)
85 mL , Volume (feed)

width

17.8 cm (7 in.)
178 mm

matrix

(Ultracel)

pore size

3 kDa pore size

pH

2-13

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Pellicon® 3 cassettes with Ultracel® membrane are advanced, high-performance cassettes that are ideal for today’s higher titer therapeutic antibodies, as well as for the more demanding filtration processes that require higher operating pressures, temperatures, concentrations and caustic cleaning regimes.
From small-scale to full-scale production, Pellicon® 3 cassettes are designed for use in research, process scale-up/scale-down, applications development and full-scale manufacturing. The Pellicon® 3 cassette design and automated manufacturing process provides unbeatable performance consistency and enhanced linear scalability between cassette sizes. Pellicon® 3 cassettes also offer greater cassette size selection for improved scale-up and scale-down process development. The streamlined design allows operators to quickly and easily handle, install and remove Pellicon® 3 cassettes. The materials of construction are compatible with a broad range of chemical cleaning agents that many TFF systems require to ensure proper sanitization.

Application

monoclonal antibodies, recombinant and non-recombinant proteins, vaccines

Features and Benefits

  • Optimum product recovery using proven composite membrane technology
  • Fast, reliable scale-up/-down from lab to production scale
  • Rugged, reliable design ideally suited to filtration processes with higher operating pressures, temperatures and caustic cleaning regimes
  • Automated manufacturing delivers unbeatable performance consistency and reliability
  • Easy to install and clean
  • Extreme temperature and chemical compatibility
  • Choice of screens to best optimize your process

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service