Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P2PLACV01

Millipore

Pellicon® 2 Mini Ultrafiltration Module

filtration area 0.1 m2, NMWCO 1 kDa

Synonym(s):

Pellicon 2 Mini Ultrafiltration Module PLAC V 0.1 m2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806

material

polypropylene screen
polyurethane adhesive
regenerated cellulose

Quality Level

manufacturer/tradename

Pellicon®

parameter

3.5 bar max. transmembrane pressure (50 psig) at 30 °C
5-35 L/min recirculation rate at 0.4 bar (6 psi)

technique(s)

analytical sample preparation: suitable
protein purification: suitable

H

2.16 cm (0.85 in.)

L

21 cm (8.3 in.)

W

5.6 cm (2.2 in.)

filtration area

0.1 m2

membrane nominal area

0.1 m2

gravimetric extractables

≤1500 mg/m2

matrix

Ultracel®

pore size

1 kDa NMWCO

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Mini Cassette
Filter Code: PLAC
Filter Type: Ultrafiltration

Features and Benefits

Type V screen (open channel)

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.
Prefiltration Rquirements: 100 µm

Analysis Note

Gravimetric Extractables: after a water flush

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service