Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN1802

Sigma-Aldrich

Anti-pan-PMCA Antibody, clone 5F10

clone 5F10, from mouse

Synonym(s):

Plasma membrane calcium-transporting ATPase, Plasma membrane calcium pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F10, monoclonal

species reactivity

chicken, rat, human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Plasma membrane calcium-transporting ATPase (isoforms 1-4) UniProt: P20020, Q01814, Q16720, P23634; also known as PMCA, Plasma membrane calcium pump) is encoded by the ATP2B1, ATP2B2, ATP2B3, ATP2B4 genes (Gene ID: 490; 491; 492; 493) in human. PMCAs are multi-pass membrane proteins that act as a calcium/calmodulin-regulated and magnesium-dependent enzymes and catalyze the hydrolysis of ATP coupled with the transport of calcium out of the cell. PMCAs serve as the main off switch for calcium signals in all types of cells. Although there are four separate genes that code for PMCAs 1-4, alternative splicing of the primary transcripts further increases the number of PMCA variants to about 20, which differ significantly in their regulatory and kinetic properties. PMCA activity is shown to be regulated at (a) the transcriptional level (b) by calmodulin binding and (c) by their subcellular localization. The localization of different PMCAs is regulated in various complex, cell type-specific ways. PMCA1 is ubiquitously expressed, PMCA2 is mainly expressed in brain cortex, but lower levels are also found in skeletal muscle, heart muscle, stomach, liver, kidney and lung. PMCA3 is highly expressed in the cerebellum, particularly in the presynaptic terminals of parallel fibers-Purkinje neurons. PMCA4 is expressed ubiquitously, however, its tissue distribution varies with its isoforms. (Ref.: Antalffy, G et al. (2013). Biochim. Biophys. Acta 1833(12); 2561-2572; Strehler, EE (2015). Biochem. Biophys. Commun. 460 (1); 26-33)

Specificity

Clone 5F10 detects all isoforms of Plasma membrane calcium-transporting ATPase (PMCA) in human, mouse, and rat.

Immunogen

Purified human erythrocyte calcium ATPase.

Application

Anti-pan-PMCA, clone 5F10, Cat. No. MABN1802, is a mouse monoclonal antibody that targets pan-Plasma membrane calcium-transporting ATPase (PMCA) and has been tested in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected pan-PMCA in human pancreas and human kidney tissue.

Immunocytochemistry Analysis: A representative lot detected pan-PMCA in Immunocytochemistry applications (Paszty, K., et. al. (2005). Biochem J. 391(Pt 3):687-92; DeMarco, S.J., et. al. (2002) J Biol Chem. 277(12):10506-11).

Immunoprecipitation Analysis: A representative lot immunoprecipitated pan-PMCA in Immunoprecipitation applications (DeMarco, S.J., et. al. (2002) J Biol Chem. 277(12):10506-11).

Western Blotting Analysis: A representative lot detected pan-PMCA in Western Blotting applications (Paszty, K., et. al. (2005). Biochem J. 391(Pt 3):687-92; Caride, A.J., et. al. (1996). Biochem J. 316 ( Pt 1):353-9).

ELISA Analysis: A representative lot detected pan-PMCA in ELISA applications (Caride, A.J., et. al. (1996). Biochem J. 316 ( Pt 1):353-9).
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry (Paraffin) in human cerebral cortex tissue

Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected pan-PMCA in human cerebral cortex tissue sections.

Target description

134 to 139 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan J Durbin et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 43(32), 5741-5752 (2023-07-21)
Neurotransmission is shaped by extracellular pH. Alkalization enhances pH-sensitive transmitter release and receptor activation, whereas acidification inhibits these processes and can activate acid-sensitive conductances in the synaptic cleft. Previous work has shown that the synaptic cleft can either acidify because

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service