Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABE70

Sigma-Aldrich

Anti-MBNL1 Antibody, clone 4A8

clone 4A8, from mouse

Synonym(s):

muscleblind-like (Drosophila), muscleblind (Drosophila)-like, muscleblind-like protein 1, Triplet-expansion RNA-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A8, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MBNL1(4154)

General description

Muscleblind 1 (MBNL1) is a member of the muscleblind family. MBNL1 is involved in pre-mRNA alternative splicing and acts as either splicing activator or repressor on specific pre-mRNA targets. This protein contains four C3H1-type zinc fingers and binds to expanded CUG repeat RNA. High expression of MBNL1 has been observed in cardiac and skeletal muscle as well as in differentiating myoblasts. MBNL1 is known to be involved in the pathogenesis of dystrophia myotonica type 1 (DM1). For those afflicted with this muscular disorder, MBNL1 is sequestered by DMPK RNAs that contain CUG triplet repeat expansions.

Immunogen

Full length recombinant protein corresponding to human MBNL1.

Application

Anti-MBNL1 Antibody, clone 4A8 is a Mouse Monoclonal Antibody for detection of MBNL1 also known as muscleblind-like protein 1, Triplet-expansion RNA-binding protein & has been validated in WB, ICC.
Immunocytochemistry Analysis:
1:4 dilution from a representative lot detected MBNL1 in myotonic dystrophy cells. Data courtesy of Dr. Ian Holt, Wolfson Centre for Inherited Neuromuscular Disease.

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: 1:10,000 dilution of this antibody detected MBNL1 on 10 µg of HEK293 cell lysate.

Target description

~39 kDa observed

Physical form

Format: Purified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphael I Benhamou et al.
Cell chemical biology, 27(2), 223-231 (2020-01-26)
Myotonic dystrophy type 2 (DM2) is a genetically defined disease caused by a toxic expanded repeat of r(CCUG) [r(CCUG)exp], harbored in intron 1 of CCHC-type zinc-finger nucleic acid binding protein (CNBP) pre-mRNA. This r(CCUG)exp causes toxicity via a gain-of-function mechanism
Raphael I Benhamou et al.
Journal of medicinal chemistry, 63(14), 7827-7839 (2020-07-14)
RNA repeat expansions are responsible for more than 30 incurable diseases. Among them is myotonic dystrophy type 1 (DM1), the most common form of adult on-set muscular dystrophy. DM1 is caused by an r(CUG) repeat expansion [r(CUG)exp] located in the
Fan Zhang et al.
Human molecular genetics, 26(16), 3056-3068 (2017-05-24)
Myotonic dystrophy Type 1 (DM1) is a rare genetic disease caused by the expansion of CTG trinucleotide repeats ((CTG)exp) in the 3' untranslated region of the DMPK gene. The repeat transcripts sequester the RNA binding protein Muscleblind-like protein 1 (MBNL1)
Alicia J Angelbello et al.
Cell chemical biology, 28(1), 34-45 (2020-11-07)
Many diseases are caused by toxic RNA repeats. Herein, we designed a lead small molecule that binds the structure of the r(CUG) repeat expansion [r(CUG)exp] that causes myotonic dystrophy type 1 (DM1) and Fuchs endothelial corneal dystrophy (FECD) and rescues

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service