Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE331

Sigma-Aldrich

Anti-MDM2 Antibody, clone 4B2C1.11

clone 4B2C1.11, from mouse

Synonym(s):

E3 ubiquitin-protein ligase Mdm2, Double minute 2 protein, Hdm2, Oncoprotein Mdm2, p53-binding protein Mdm2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B2C1.11, monoclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MDM2(4193)
mouse ... Mdm2(17246)

General description

MDM2 is over-expressed in many tumors. Its principal function is the ubiquitination and degradation of p53 tumour suppressor protein. This monoclonal antibody also recognizes a peptide epitope around Thr-216 of murine MDM2 when Thr-216 is unphosphorylated. SMP14 also cross reacts with some cytokeratins (6, 14 & 16). This is only a problem when working with certain epithelial cells and not fibroblasts.

Immunogen

Recombinant protein corresponding to human MDM2.

Application

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated MDM2 in SV5-tagged, fusion p53-EGFP transfected H1299 cell lysates (Hjerpe, R., et al. (2010). 42(5):725-735.).

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected MDM2 in SV5-tagged, fusion p53-EGFP transfected H1299 cells (Hjerpe, R., et al. (2010). 42(5):725-735.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-MDM2 Antibody, clone 4B2C1.11 (mouse monoclonal antibody) validated in WB, IP, ICC to detect MDM2 also known as E3 ubiquitin-protein ligase Mdm2, Double minute 2 protein, Hdm2, Oncoprotein Mdm2, p53-binding protein Mdm2.

Quality

Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 0.25 µg/mL of this antibody detected MDM2 in 10 µg of mouse brain tissue lysate.

Target description

~55 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Bonacci et al.
PloS one, 12(1), e0169988-e0169988 (2017-01-19)
NUB1 (Nedd8 ultimate buster 1) is an adaptor protein which negatively regulates the ubiquitin-like protein Nedd8 as well as neddylated proteins levels through proteasomal degradation. However, molecular mechanisms underlying this function are not completely understood. Here, we report that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service