Skip to Content
MilliporeSigma

MABC612

Anti-Procadherin FAT1 Antibody, clone C257

clone C257, from mouse

Synonym(s):

Cadherin family member 7, Cadherin-related tumor suppressor homolog, Protein fat homolog

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
C257, monoclonal
Application:
IP, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C257, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FAT1(2195)

General description

>260 kDa observed. Procadherin FAT1 is an extremely large cell surface receptor with a calculated MW of 506 kDa and it′s highly glycosolated. It may be difficult to resolve the protein by western blot. An uncharacterized band at 50 kDa and other lower MWs may be observed in some lysates.
Procadherin FAT1 protein is a cell membrane bound protein that also has an active nuclear-translocation component that can alter specific gene expression and cell proliferation. Procadherin FAT1 is important in cell polarization and directed cell migration and also modulates cell-cell contact interactions. Procadherin FAT1 is expressed in epithelial, endothelial and smooth muscle cells and plays roles in their response to injury and is often highly induced after arterial injury and effects vascular smooth muscle cell growth and migration. Procadherin FAT1 is also important in cancer where it appears to function as both an oncogene and a tumor suppressor depending upon the cell context. Also recent research has shown that soluble FAT1 can be a novel biomarker candidate for such cancers as pancreatic cancer.

Immunogen

Recombinant protein corresponding to Human FAT1.

Application

Research Category
Apoptosis & Cancer
This Anti-Procadherin FAT1 Antibody, clone C257 is validated for use in Western Blotting and Immunoprecipitation for the detection of Procadherin FAT1.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Procadherin FAT1 in 10 µg of FAT1 transfected MV3 cell lysate.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Procadherin FAT1 in 10 µg of FAT1 transfected MDA-MB-231 and MDA-MB-468 cell lysate.
Immunoprecipitation Analysis: A representative lot immunoprecipitated Procadherin FAT1 in C257 cell lysate. (Courtesy of Dr. R. Thorne, U of Newcastle, AU.)

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in FAT1 transfected NIH-3T3 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Procadherin FAT1 in 10 µg of FAT1 transfected NIH-3T3 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library