Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABC151

Sigma-Aldrich

Anti-Hyaluronan synthase 2 Antibody, clone 2B11.1

clone 2B11.1, from mouse

Synonym(s):

Hyaluronan synthase 2, Hyaluronate synthase 2, Hyaluronic acid synthase 2, HA synthase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B11.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HAS2(3037)

General description

Hyaluronan synthase 2 (HAS2) is also known as Hyaluronate synthase 2, or Hyaluronic acid (HA) synthase 2. As the name suggests, HAS2 is involved in hyaluronan/hyaluronic acid synthesis. Hyaluronan synthase 2 is expressed in fibroblasts. Chromosomal aberrations involving Hyaluronan synthase 2 may be a cause of lipoblastomas, usually diagnosed in children.

Specificity

This antibody recognizes the cytoplasmic domain of Hyaluronan synthase 2.

Immunogen

Epitope: Cytoplasmic domain
GST-tagged recombinant protein corresponding to the cytoplasmic domain of human Hyaluronan synthase 2.

Application

Detect Hyaluronan synthase 2 using this mouse monoclonal antibody, Anti-Hyaluronan synthase 2 Antibody, clone 2B11.1 validated for use in western blotting & IHC (Paraffin).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected Hyaluronan synthase 2 in 10 µg of K562 cell lysate.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Hyaluronan synthase 2 in human breast adenocarcinoma cell tissue.

Quality

Evaluated by Western Blotting in MRC-5 cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Hyaluronan synthase 2 in 200 µg of MRC-5 cell lysate.

Target description

~ 64 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MRC-5 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service