Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1123

Sigma-Aldrich

Anti-CEACAM5 (CD66e) Antibody, clone M5A

clone M5A, from mouse

Synonym(s):

Carcinoembryonic antigen-related cell adhesion molecule 5, Carcinoembryonic antigen, Meconium antigen 100, CD66e

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

M5A, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CEACAM5(1048)

General description

Carcinoembryonic antigen-related cell adhesion molecule 5 (UniProt: P06731; also known as Carcinoembryonic antigen, CEA, Meconium antigen 100, CD66e) is encoded by the CEACAM5 (also known as CEA) gene (Gene ID: 1048) in human. CEA is a cell surface heavily glycosylated homodimeric glycoprotein that plays a role in cell adhesion and in intracellular signaling. CEA molecules can either be transmembrane or GPI-linked and are differentially expressed between species. CEA is restricted to the apical face of intestinal epithelial cells in the adult, but it has more diffuse distribution during embryonic development and in tumors. It functions as a calcium independent adhesion molecule through homophilic and heterophilic interactions with CEACAM-1. CEA participates in diverse physiological processes, including cell adhesion, differentiation, proliferation, and survival, as well as in carcinogenesis and bacterial pathogenesis. CEA molecules from neighboring cells can interact via their respective extracellular N-terminal IgV-like domain and mediate cell-cell adhesion through trans-oligomerization. CEA is synthesized with a signal peptide (aa 1-34) and a propeptide region (aa 686-702) that are cleaved to produce the mature form. It has seven Ig-like domains. Two isoforms of CEA have been reported that are produced by alternative splicing. Clone M5A is a humanized monoclonal antibody that displays high specificity and sub-nanomolar affinity for tumor-associated CEA. (Ref. Yazaki, PJ et al., (2004). Protein Eng. Des Sel. 17(5);481-489.

Specificity

Clone M5A is a humanized monoclonal antibody that detects human Carcinoembryonic antigen-related cell adhesion molecule 5 (CEA). Note: please use anti-human secondary antibody for detedtion.

Immunogen

Full length purified human Carcinoembryonic antigen-related cell adhesion molecule 5 (CEA).

Application

Anti-CEACAM5 (CD66e), clone M5A, Cat. No. MABC1123, is a humanized monoclonal antibody that detects Carcinoembryonic antigen-related cell adhesion molecule 5 and has been tested for use in Flow Cytometry, Immunohistochemistry, and Radioimmunoassay.
Immunohistochemistry Analysis: A representative lot detected CEACAM5 (CD66e) in Immunohistochemistry applications (Nittka, S., et. al. (2014). PLoS One. 9(9):e106921).

Radioimmunoassay Analysis: A representative lot detected CEACAM5 (CD66e) in Radioimmunoassay applications (Nittka, S., et. al. (2014). PLoS One. 9(9):e106921; Yazaki, P.J., et. al. (2004). Protein Eng Des Sel. 17(5):481-9; Yazaki, P.J., et. al. (2013). Protein Eng Des Sel. 26(3):187-93).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Flow Cytometry in HT29 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected CEACAM5 (CD66e) in one million HT29 cells.

Target description

76.80 kDa calculated.

Physical form

Format: Purified
Protein A purified
Purified humanized monoclonal antibody in PBS with 0.05% sodium azide. This humanized antibody was produced by transfecting the cDNA into murine myeloma NSO host cells for stable expression.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service