Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1111

Sigma-Aldrich

Anti-Angiogenin Antibody, clone C527

clone C527, from mouse

Synonym(s):

Angiogenin-1, Ribonuclease 5, RNase 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

C527, monoclonal

species reactivity

mouse

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

Gene Information

mouse ... Ang(11727)

General description

Angiogenin (UniProt: P21570; also known as EC: 3.3.27.-, Angiogenin-1, Ribonuclease 5, RNase 5) is encoded by the Ang (also known as Ang1, Rnase5, Rnase5a) gene (Gene ID: 11727) in murine species. Angiogenin is a homodimeric protein that belongs to the pancreatic ribonuclease family. It is synthesized with a signal peptide (aa 1-24), which is subsequently cleaved off. Angiogenin binds to actin on the surface of endothelial cells and is rapidly endocytosed and translocated to the nucleus where it accumulates in the nucleolus and binds to DNA. Its nuclear localization signal is located in amino acids 55-59. Angiogenin induces vascularization of normal and malignant tissues and its activity is regulated by interaction with RNH1 in vivo. The growth stimulatory functions of Angiogenin are mediated through rRNA transcription. It stimulates ribosomal RNA synthesis including that containing the initiation site sequences of 45S rRNA. It cleaves tRNA within anticodon loops to produce tRNA-derived stress-induced fragments (tiRNAs), which inhibit protein synthesis and triggers the assembly of stress granules (SGs). Under stress, Angiogenin is translocated to SGs and mediates the production of tRNA-derived stress-induced small RNA (tiRNA); which enhance cellular survival by simultaneously suppressing global protein translation, saving anabolic energy, and permitting internal ribosomal entry sequence (IRES)-mediated protein translation of anti-apoptotic genes.

Specificity

Clone C527 detects Angiogenin in murine cells.

Immunogen

Full length recombinant murine Angiogenin.

Application

Anti-Angiogenin, clone C527, Cat. No. MABC1111, is a mouse monoclonal antibody that detects Angiogenin and has been tested for use in ELISA, Immunofluorescence, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected Angiogenin in Immunofluorescence applications (Goncalves, K.A., et. al. (2016). Cell. 166(4):894-906).

ELISA Analysis: A representative lot detected Angiogenin in ELISA applications (Goncalves, K.A., et. al. (2016). Cell. 166(4):894-906).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting with recombinant mouse angiogenin.

Western Blotting Analysis: 4 µg/mL of this antibody detected recombinant mouse angiogenin.

Target description

~16 kDa observed; 16.23 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service