Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB4148

Sigma-Aldrich

Anti-MRP2 Antibody, clone M2 I-4

culture supernatant, clone M2 I-4, Chemicon®

Synonym(s):

cMOAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

M2 I-4, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... ABCC2(1244)

Specificity

This antibody reacts with an internal epitope of MRP2, a 190-200 kDa transmembrane protein previously known as the canalicular multi-organic anion transporter (cMOAT). MRP2 is a member of the MRP family of multidrug resistance related proteins, and MRP2 overexpression has been observed in a subset of cisplatin resistant cell lines. This antibody does not cross-react with the human MDR1, MRP1, MRP3 or MRP5 gene products.

Immunogen

Fusion protein of MRP2 containing aa 215-310 of the protein.

Application

This Anti-MRP2 Antibody, clone M2 I-4 is validated for use in WB, IC, IH for the detection of MRP2.
Western blotting: 1:20 - 1:50 with anti-mouse HRP

Immunocytochemistry: 1:20 - 1:50 on acetone fixed cytospin preparations

Immunohistochemistry: 1:20 on acetone fixed frozen tissue sections

Optimal working dilutions must be determined by end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Päivi Myllynen et al.
Toxicology and applied pharmacology, 232(2), 210-217 (2008-08-06)
We have studied the role of ATP binding cassette (ABC) transporters in fetal exposure to carcinogens using 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) a known substrate for ABC transporters as a model compound. In perfusion of human term placenta, transfer of (14)C-PhIP (2 microM)
Michael J Hafey et al.
Drug metabolism and disposition: the biological fate of chemicals, 48(11), 1147-1160 (2020-09-19)
Hepatocellular accumulation of bile salts by inhibition of bile salt export pump (BSEP/ABCB11) may result in cholestasis and is one proposed mechanism of drug-induced liver injury (DILI). To understand the relationship between BSEP inhibition and DILI, we evaluated 64 DILI-positive
Tobacco carcinogen NNK transporter MRP2 regulates CFTR function in lung epithelia: implications for lung cancer.
Chunying Li,John D Schuetz,Anjaparavanda P Naren
Cancer letters null
M Kummu et al.
Placenta, 33(10), 859-865 (2012-08-16)
Heavy metals such as cadmium, lead and methylmercury are known to be neurotoxic to developing fetus. ABCG2 which is an efflux transporter located in the maternal facing membranes of human placenta protects fetus from xenobiotics by transferring compounds from syncytiotrophoblast
C C Paulusma et al.
Science (New York, N.Y.), 271(5252), 1126-1128 (1996-02-23)
The human Dubin-Johnson syndrome and its animal model, the TR(-) rat, are characterized by a chronic conjugated hyperbilirubinemia. TR(-) rats are defective in the canalicular multispecific organic anion transporter (cMOAT), which mediates hepatobiliary excretion of numerous organic anions. The complementary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service