Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB3086

Sigma-Aldrich

Anti-Ryanodine Receptor Antibody, clone RYR.1

clone RYR.1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RYR.1, monoclonal

species reactivity

rat, human, mouse, pig, bovine, canine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RYR1(6261)

Specificity

Reacts with the C-terminus cytoplasmic domain of ryanodine receptor. Ryanodine receptors have been shown to play critical roles in the intracellular Ca2+ signaling occurring during cell activation in muscle cells and non-muscle cells. MAB3086 reacts with ryanodine receptor (MW approx. 500kDa) isolated from a variety of cell types (e.g. lymphocytes, macrophages, granulocytes, fibroblasts, epithelial, endothelial cells, skeletal muscle, cardiac muscle and brain tissues).

Immunogen

Short synthetic polypeptide corresponding to the c-terminal domain of the ryanodine receptor.

Application

Anti-Ryanodine Receptor Antibody, clone RYR.1 detects level of Ryanodine Receptor & has been published & validated for use in ELISA, FC, IC, IH, IP, RIA & WB.
Immunoblot: 1:100 using enhanced chemiluminescence detection. It is suggested that you perform a dot blot to verify presence of the RyR protein if you are having difficulties performing an immunoblot assay.

Immunocytochemistry: 1:100 by indirect immunofluorescence. Visualization by confocal microscopy is required, as detection by standard fluorescent microscopy will not be adequate to detect the RyR. Additionally, fluorescent, not enzymatic, detection is required. Due to the intensity of confocal lasers, use of an anti-fading agent, such as DABCO, is strongly recommended.

Immunoprecipitation: 1:100

Flow cytometry: 1:500

ELISA: 1:1000

Immunohistochemistry: 1:100

RIA: 1:5000

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Signaling Neuroscience

Physical form

Format: Purified
Purified immunoglobulin from culture supernatant. In 10 mM sodium phosphate, 150 mM NaCl, 0.01% NaN3, pH. 7.5

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chantal Donovan et al.
American journal of respiratory cell and molecular biology, 53(4), 471-478 (2015-02-19)
Small airways are a major site of airflow limitation in chronic obstructive pulmonary disease (COPD). Despite the detrimental effects of long-term smoking in COPD, the effects of acute cigarette smoke (CS) exposure on small airway reactivity have not been fully

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service