Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB2613

Sigma-Aldrich

Anti-UPAR-Associated Protein Antibody, clone 2.h.9:F12

clone 2.h.9:F12, from mouse

Synonym(s):

CD280, Endocytic receptor 180, Macrophage mannose receptor 2, UPAR-associated protein, Urokinase receptor-associated protein, Endocytic receptor 180, C-type mannose receptor 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2.h.9:F12, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MRC2(9902)

General description

Urokinase plasminogen activator receptor-associated protein (uPARAP/Endo180/MRC2) is a member of the macrophage mannose receptor family of endocytic transmembrane glycoproteins. UPAR-Associated Protein is a membrane protein involved in matrix turnover during tissue remodeling, particularly the uptake and intracellular degradation of collagen, (Sulek, 2007).

Specificity

The antibody recognizes UPAR-Associated Protein.

Immunogen

uPAR D3-tagged recombinant protein corresponding to human UPAR-Associated Protein.

Application

Anti-UPAR-Associated Protein Antibody, clone 2.h.9:F12 is an antibody against UPAR-Associated Protein for use in WB, IH.

Quality

Evaluated by Western Blot in U937 cell lysate.

Western Blot Analysis: 0.2 µg/ml of this antibody detected UPAR-Associated Protein on 10 µg of U937 cell lysate.

Target description

~ 180 kDa

Physical form

Format: Purified

Analysis Note

Control
U937 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service