Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1645

Sigma-Aldrich

Anti-Dystrophin Antibody, clone 1808

clone 1808, Chemicon®, from mouse

Synonym(s):

Anti-BMD, Anti-CMD3B, Anti-DXS142, Anti-DXS164, Anti-DXS206, Anti-DXS230, Anti-DXS239, Anti-DXS268, Anti-DXS269, Anti-DXS270, Anti-DXS272, Anti-MRX85

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1808, monoclonal

species reactivity

chicken, mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DMD(1756)

Specificity

By Western blot MAB1645 recognizes a single protein having a relative molecular weight of 300 kD. Reacts to mammalian skeletal muscle dystrophin. Shows no staining of mdx mouse muscle. Labels normal human muscle but not Duchenne muscle.

STAINING PATTERN:

On cryostat sections of normal mouse diaphragm MAB1645 gives strong immunofluorescence staining of the entire sarcolemma, with particularly strong staining of neuromuscular junctions.

SPECIES REACTIVITY:

Labels skeletal muscle from rat, chicken and Xenopus laevis. Labels cardiac muscle from rat and normal, but not mdx, mouse. Also labels smooth muscle from chicken gizzard.

Immunogen

Peripheral membrane proteins extracted from Torpedo postsynaptic membranes.

Application

Anti-Dystrophin Antibody, clone 1808 detects level of Dystrophin & has been published & validated for use in WB, IH.
Western blot

Immunohistochemistry

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Liquid in 0.02 M phosphate buffer (pH 7.6), 250 mM NaCl with 0.1% sodium azide.

Storage and Stability

Maintain at -20ºC for up to 12 months in convenient aliquots. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Payam Soltanzadeh et al.
Neuromuscular disorders : NMD, 20(8), 499-504 (2010-07-16)
Manifesting carriers of DMD gene mutations may present diagnostic challenges, particularly in the absence of a family history of dystrophinopathy. We review the clinical and genetic features in 15 manifesting carriers identified among 860 subjects within the United Dystrophinopathy Project
Andreas Schaefer et al.
PloS one, 11(2), e0148259-e0148259 (2016-02-04)
Previous small animal models for simulation of mechanical unloading are solely performed in healthy or infarcted hearts, not representing the pathophysiology of hypertrophic and dilated hearts emerging in heart failure patients. In this article, we present a new and economic
Andreas Schaefer et al.
Scientific reports, 9(1), 5710-5710 (2019-04-07)
Mechanical unloading (MU) by implantation of left ventricular assist devices (LVAD) has become clinical routine. This procedure has been shown to reverse cardiac pathological remodeling, with the underlying molecular mechanisms incompletely understood. Most studies thus far were performed in non-standardized
Mariana Fernandez-Caggiano et al.
Nature metabolism, 2(11), 1223-1231 (2020-10-28)
Cardiomyocytes rely on metabolic substrates, not only to fuel cardiac output, but also for growth and remodelling during stress. Here we show that mitochondrial pyruvate carrier (MPC) abundance mediates pathological cardiac hypertrophy. MPC abundance was reduced in failing hypertrophic human
Tessa R Werner et al.
Scientific reports, 9(1), 11494-11494 (2019-08-09)
Afterload enhancement (AE) of rat engineered heart tissue (EHT) in vitro leads to a multitude of changes that in vivo are referred to as pathological cardiac hypertrophy: e.g., cardiomyocyte hypertrophy, contractile dysfunction, reactivation of fetal genes and fibrotic changes. Moreover

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service