Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB10762

Sigma-Aldrich

Anti-Transthyretin Antibody, clone 7G8-E11-E7

clone 7G8-E11-E7, from mouse

Synonym(s):

prealbumin, amyloidosis type I, thyroxine-binding prealbumin, TBPA, TTR, ATTR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7G8-E11-E7, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TTR(7276)

General description

Transthyretin is one of the three major thyroid hormone-binding proteins in plasma and/or cerebrospinal fluid of vertebrates. It exists mainly as a homotetrameric protein of approximately 55 kDa in plasma and, in sera and cerebrospinal fluid, is a carrier of the thyroid hormone thyroxine (T4) and retinol. While mostly known for being the transporter of thyroxine and retinol, when mutated, TTR is also well-described as the cause of familial amyloid polyneuropathy, a neurodegenerative lethal disorder characterized by systemic deposition of TTR amyloid fibrils, particularly in the peripheral nervous system. Recently, studies have shown that TTR has an important function development of peripheral and central nervous system physiology, namely by participating in behavior, in the maintenance of normal cognitive processes during ageing, amidated neuropeptide processing and nerve regeneration. Additionally, it has been proposed that TTR is neuroprotective in Alzheimer′s disease, by preventing the formation of amyloid beta fibrils (Fleming C.E. et al. (2009) Prog Neurobiol., Epub ahead of print

Specificity

This antibody recognizes Transthyretin.

Immunogen

Purified human Transthyretin.

Application

This Anti-Transthyretin Antibody, clone 7G8-E11-E7 is validated for use in ELISA, WB for the detection of Transthyretin.

Quality

Evaluated by Western Blot in human placenta lysate.

Western Blot Analysis: 2 µg/ml of this antibody detected Transthyretin in 10 µg of human placenta lysate.

Target description

Transthyretin exists mainly as a homotetrameric protein of approximately 55 kDa in plasma. A single band of ~26 kDa was observed, which is an approximate size of a dimeric component that makes up the said homotetramer.

Physical form

Format: Purified

Analysis Note

Control
Human placenta lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service