Skip to Content
MilliporeSigma

IPAKMET00

IPAK Meta® Polishing Cartridge for Purified Water

greener alternative

For Milli-Q® IQ/EQ 7000 and EQ 7008/16 ultrapure water systems fed with purified water. This product replaces IPAKMETA1.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104212
NACRES:
NB.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

packaging

pkg of 1 unit

manufacturer/tradename

Milli-Q®

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

humidity measuring range

10-80% (In original sealed packaging to protect purification elements)

H

39.5 cm (15.5 in.)

compatibility

for use with Milli-Q® EQ 7000, for use with Milli-Q® EQ 7008, for use with Milli-Q® EQ 7016, for use with Milli-Q® IQ 7000

greener alternative category

shipped in

ambient

storage temp.

2-30°C

General description

This IPAK Meta® polishing cartridge for purified water contains a combination of Jetpore® mixed-bed ion-exchange resin and innovative IQnano ion-exchange media to remove trace ions from pure water.

Combined with an IPAK Quanta® water polishing cartridge for purified water (IPAKQUA00), it enables the production of ultrapure (Type 1) water for sensitive laboratory applications.
We continue to demonstrate our commitment to a sustainable future by exploring ways to improve existing products in our portfolio. We have reviewed our purification cartridges to identify key opportunities to minimize impact without compromising on the product performance. This product has benefited from an optimization of the resin mix, the packaging and the end-of-life management. Therefore, it is identified as a greener alternative product in the category reengineered. View the DfS scorecard for the new IPAK Meta® cartridge for pure water.

Application

  • For Milli-Q® IQ/EQ 7000 and EQ 7008/16 ultrapure water purification systems fed with purified water
  • Delivers ultrapure (Type 1) water for sensitive applications

Features and Benefits

  • e-Sure tag enables RFID connection with the system, for seamless consumable status monitoring, data management and full traceability
  • Supplied with a Drain Cap, to remove excess water and reduce the weight of the exhausted cartridge at disposal
  • 78% of the parts suppliers are ISO 14001 environmental management system-certified

Legal Information

IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
JETPORE is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library