Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HAGE1MAG-20K

Millipore

MILLIPLEX® Human Aging Magnetic Bead Panel 1 - Metabolism Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

species reactivity

human

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Aging associated physiological changes and the pathological consequences of these changes make the elderly population vulnerable to many chronic diseases, including diabetes, cancers, cardiovascular and neurodegenerative diseases, etc. The complexity of the aging process has stimulated intensive research over the years. Aberrant glucose metabolism, elevated chronic inflammatory levels, progressive cellular senescence and other pathophysiological conditions have proven to be associated with the aging process.The MILLIPLEX® MAP Human Aging Panel 1 covers multifaceted biomarkers of aging-related conditions. Monitoring these novel biomarkers would help researchers to understand the basic mechanisms that drive aging and eventually lead to improved clinical interventions for extending the human health span.The MILLIPLEX® MAP Human Aging Panel 1 is an 10-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum, plasma, and in vitro samples: C-C motif chemokine 27 (CTACK/CCL27), Fibroblast growth factor 21 (FGF-21), Growth/differentiation factor 11 (GDF-11), Growth/differentiation factor 15 (GDF-15), Gonadoliberin (GnRH), Interleukin-6 (IL-6), Interleukin-10 (IL-10), Interleukin 18 (IL-18), Jagged 1 protein (Jag1), and Leptin. Note: IL-10 is recommended for in vitro sample use only; do not test IL-10 in serum or plasma samples.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

For optimal sensitivity, use an overnight (16-18 hour) incubation. A two hour incubation can also be used. Serum and plasma samples require (recommend) a 1:2 dilution with Assay Buffer. Tissue culture supernatants may be run neat or appropriately diluted in control medium. Each well requires 25 μL of sample, standard or control. Analytes available:CTACK/CCL27, FGF-21, GDF-11, GDF-15, Gonadoliberin (GnRH), IL-6, IL-10, IL-18, Jagged 1 protein (Jag1), & Leptin

Features and Benefits

Configuration: Premixed

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Multiplex immunoassays like MILLIPLEX® Human Myokine Panel enable comprehensive myokine analysis, providing insights into related diseases.

Multiplex immunoassays like MILLIPLEX® Human Myokine Panel enable comprehensive myokine analysis, providing insights into related diseases.

Multiplex immunoassays like MILLIPLEX® Human Myokine Panel enable comprehensive myokine analysis, providing insights into related diseases.

Multiplex immunoassays like MILLIPLEX® Human Myokine Panel enable comprehensive myokine analysis, providing insights into related diseases.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service