Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EZRBDG-110K

Sigma-Aldrich

αnti-RBD IgG Human ELISA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

grade

for protein analysis

clone

monoclonal

feature

accuracy 98% Av

species reactivity

human

description

3.12-100 ng/mL (standard curve range)

technique(s)

ELISA: suitable

detection method

colorimetric

General description

The Receptor Binding Domain (RBD) protein is a subunit of the S1 Spike protein of SARS-CoV-2. Studies indicate that it is strongly immunogenic and can elicit a strong humoral response with rises in titer of specific antibodies post-infection with the virus. IgG antibodies specific to RBD have been demonstrated in several studies as playing a meaningful role in the study of SARS-CoV-2 disease progression.
For Research Use Only. Not For Use In Diagnostic Procedures.

Application

2 hour 30 minute assay, room temperature, a starting 1:100 dilution of human serum or plasma recommended .

Analysis Note

Serum Linearity: 100% Av Plasma Linearity: 98% Av

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system, Respiratory Tract

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service