Skip to Content
MilliporeSigma
All Photos(1)

Documents

EZABNZ-180K

Millipore

Advanced Benzonase ELISA

new

Synonym(s):

Endonuclease from Serratia marcescens, ELISA for the detection of Endonuclease A from Serriatta marcescens

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12161503
NACRES:
NA.84

biological source

Serratia marcescens

Quality Level

description

96 well

parameter

50 μL

assay range

intra-assay cv: <15%
standard curve range: 15.6-1000 pg/mL
inter-assay cv: <15%

technique(s)

ELISA: suitable

input

cell culture supernatant

detection method

colorimetric

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

ELISA for the specific detection of Benzonase® Nuclease (Endonuclease A from S.marcescens).

Application

96 stripwell formatt ELISA
Used for the removal of nucleic acid from protein samples.

Biochem/physiol Actions

Digests native or heat-denatured DNA and RNA.

Features and Benefits

Detects Benzonase® Nuclease using a pair of specifc monoclonal antibodies in a capture ELISA formatt. Results are determined spectrophotometrically using a Benzonase® standard curve. The assay range is 15.6-1000pg ml, with an LLOQ of 15.6pg (%CV/%Recovery <20%).

Legal Information

Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service