MilliporeSigma
Skip To
EXTSV00A1
External Solenoid valve
Regulates feed water flow, for use with Milli-Q® IQ/IX systems
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Estimated to ship TODAY
$504.00
About This Item
NACRES:
JA.13
UNSPSC Code:
41104200
$504.00
Estimated to ship TODAYDetails
Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015
General description
The external solenoid valve stops feed water at the source in case a leak is detected to help prevent flooding.
The valve stops feed water to Milli-Q® IQ 7003/05/10/15 or Milli-Q® IX 7003/05/10/15 water purification systems.
Application
When connected to a leak detector/water sensor, this external solenoid valve prevents flooding.
Features and Benefits
Protects the lab from water damage.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
ZLXL00ESVZFDISTPA1MPGP002A1
compatibility
for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7010, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7015, for use with Milli-Q® IX 7005, for use with Milli-Q® IQ 7005
compatibility
for use with Milli-Q® CLX, for use with Milli-Q® HR, for use with Milli-Q® HX
compatibility
for use with Milli-Q® EQ 7000, for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015
compatibility
for use with Milli-Q® EQ 7000, for use with Milli-Q® EQ 7008, for use with Milli-Q® EQ 7016, for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7000, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015
packaging
pkg of 1 unit
packaging
pkg of 1 unit
packaging
-
packaging
pkg of 1 unit
Still not finding the right product?
Explore all of our products under External Solenoid valve