Skip to Content
MilliporeSigma
All Photos(1)

Documents

DX0835

Supelco

Dichloromethane

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Methylene Chloride, Dichloromethane, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-838-9

Agency

meets USP/NF

Quality Level

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

1223 °F

expl. lim.

22 %

impurities

≤0.0003 meq/g Titrable acid
≤0.02% Water (H2O)

evapn. residue

≤0.002%

color

APHA: ≤10

refractive index

n20/D 1.424 (lit.)

pH

7 (20 °C)

bp

39.8-40 °C (lit.)

mp

-95 °C
−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DX0835P-4 is packaged in poly coated glass bottles
Stabilized with an alkene.
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Appearance: Clear Liquid
Assay (GC): 99.5% min
Color (APHA): 10 max
Evaporation residue: 0.002% max
Free halogens: To pass test
Infrared Spectrum: Conforms to standard
Titrable acid: 0.0003meq/g max
Water (H2O): 0.02% max
Evaporation Number: 1.9

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service