Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

DDAC00010

Millipore

Direct Detect® Assay-free Cards

50 Membrane cards used for protein quantitation in the Direct Detect® Infrared Spectrometer.

Synonym(s):

Assay cards

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.22

Quality Level

manufacturer/tradename

Direct Detect®

technique(s)

protein quantification: suitable

shipped in

ambient

General description

Quantity Options:
DDAC00010-GR = 50 Cards
DDAC00010-4P = 200 Cards
DDAC00010-8P = 400 Cards

The Direct Detect Assay-free Cards are desgined for use in the the Direct Detect Spectrometer. The card contains four hydrophilic polytetrafluoroethylene (PTFE) membrane positions sized for easy sample application and surrounded by a hydrophobic ring to retain analyzed sample within the IR beam. A 2 μL sample volume is recommended in order to assure proper sample application, drying, and measurement.

The Assay-free card can be inserted into the system and analyzed immediately after preparation. The Direct Detect spectrometer has a built in drying system that will automatically dry the sample to the point where it will give an accurate reading.
The Direct Detect Assay-free card is intended for a one-time protein application, however, already prepared cards can be stored and reread later.

Legal Information

DIRECT DETECT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn how total protein quantitation with an infrared spectrometer may be a more accurate and universal alternative to Bradford, BCA, or UV-based methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service