Skip to Content
MilliporeSigma

Skip To

ABS44

Anti-MAPK 1/2 Antibody

from rabbit

Synonym(s):

ERK-1, ERT2, Extracellular signal-regulated kinase 1, Insulin-stimulated MAP2 kinase, MAP kinase 3, MAP kinase isoform p44, MAPK 3, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 3, MNK1, p44-ERK1, p44-MAPK, ERK-2, ERT1, Extrac

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
$444.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IP, WB
Citations:
15

$444.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB06182

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

species reactivity (predicted by homology)

canine (based on 94% sequence homology), bovine (based on 94% sequence homology)

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAPK1(5594)

General description

Mitogen-activated protein kinase 3 (EC 2.7.11.24; UniProt P21708; also known as ERK-1, ERT2, Extracellular signal-regulated kinase 1, Insulin-stimulated MAP2 kinase, MAP kinase 3, MAP kinase isoform p44, MAPK 3, Microtubule-associated protein 2 kinase, MNK1, p44-ERK1, p44-MAPK) is encoded by the Mapk3 (also known as Erk1, Prkm3) gene (Gene ID 50689) in rat species. ERK1 (a.k.a. MAPK 3) and ERK2 (a.k.a. MAPK 1 or MAPK 2) are related serine/threonine kinases of the Ras-Raf-MEK-ERK signaling pathway. ERK1/2 are activated by MEK1/2-catalyzed phosphorylation at their conserved Thr-Glu-Tyr (TEY) dual phosphorylation motif, first at the Tyr residue (human/rat ERK1 Tyr204/Tyr203, human/rat ERK2 Tyr187/Tyr183) and then at the Thr residue (human/rat ERK1 Thr202/Thr201, human/rat ERK2 Thr185/Thr181). ERK1/2 are proline-directed kinases that preferentially catalyze the phosphorylation of Pro-Xxx-Ser/Thr-Pro sequence motif in hundreds of cytoplasmic and nuclear substrates, including transcription factors such as Ets, Elk, and c-Fos. Besides this primary structure requirement, many ERK1/2 substrates possess a D-docking site and/or an F-docking site. A variety of scaffold proteins, including KSR1/2, IQGAP1, MP1, and β-Arrestin1/2, also participate in the regulation of ERK1/2 MAP kinase cascade. Ras-Raf-MEK-ERK signaling activity is upregulated in about one-third of all human cancers, and targeted inhibition against components of this signaling pathway represents a popular anticancer strategy.
~44/42 kDa observed. 41.26/36.30 kDa (human MAPK1 isoform 1/2), 41.14 kDa (rat MAPK1), 41.14 kDa (mouse MAPK1), 43.00/38.14 kDa (human MAPK3 isoform 1/2), 42.95/45.64 kDa (rat MAPK3 isoform 1/2), 42.93 kDa (mouse MAPK3) calculated (Met1 removed). Target sequence is not present in human MAPK3 spliced isoform 3 (ERK1b).

Immunogen

Epitope: C-terminal
KLH-conjugated linear peptide corresponding to the C-terminal sequence of rat MAPK 3 (ERK-1).

Application

Detect MAPK 3/MAPK 1 (ERK1/ERK2) using this Anti-MAPK 1/2 Antibody validated for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Signaling
Research Sub Category
MAP Kinases
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected MAPK 3/MAPK 1 (ERK1/ERK2) in BALB/3T3, A431, and L6 cell lysates.



Western Blotting (SNAP ID) Analysis: 1-2 µg/mL from a representative lot detected MAPK 3/MAPK 1 (ERK1/ERK2) in BALB/3T3, A431, and L6 cell lysates.



Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected MAPK 3/MAPK 1 (ERK1/ERK2) in NIH/3T3, A431, HeLa, and C2C12 cells.

Biochem/physiol Actions

This antibody recognizes MAPK 3/MAPK 1 (ERK1/ERK2) C-terminal sequence.

Physical form

Format: Purified
Protein A purified.
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate
Evaluated by Western Blotting in A431 cell lysate.



Western Blotting Analysis: 0.5 µg/mL of this antibody detected MAPK 3/MAPK 1 (ERK1/ERK2) in 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 06-182

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5700045M3807FCABS165A6
Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

conjugate

ALEXA FLUOR 647

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

purified from hybridoma cell culture

antibody form

purified antibody

Gene Information

human ... MAPK1(5594)

Gene Information

human ... MAPK1(5594)

Gene Information

human ... MAPK1(5594), MAPK3(5595)
rat ... Mapk1(116590), Mapk3(50689)

Gene Information

human ... MAPK1(5594)

technique(s)

immunocytochemistry: suitable, western blot: suitable, immunoprecipitation (IP): suitable

technique(s)

immunohistochemistry: 1:50-1:100, western blot: 1:500-1:2000

technique(s)

immunocytochemistry: suitable, indirect ELISA: suitable, microarray: suitable, western blot: 5-25 μg/mL using rat brain extract

technique(s)

flow cytometry: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters