Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

ABS2129

Sigma-Aldrich

Anti-IP3R 1

from rabbit

Synonym(s):

Inositol 1, 4, 5-trisphosphate receptor type I, IP3 receptor isoform 1, IP-3-R1, InsP3R1, Type 1 inositol 1, 4, 5-trisphosphate receptor, Type 1 InsP3 receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, bovine, mouse, chicken

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ITPR1(3708)
rat ... Itpr1(25262)

General description

Inositol 1,4,5-trisphosphate receptor type 1 (UniProt: P29994; also known as IP3 receptor isoform 1, IP-3-R, IP3R 1, InsP3R1, Type 1 inositol 1,4,5-trisphosphate receptor, Type 1 InsP3 receptor) is encoded by the Itpr1 (also known as Insp3r) gene (Gene ID: 25262) in rat. InsP3R1 is a multi-pass membrane protein located on endoplasmic reticulum (ER) membrane. Its highest levels are found in neurons, with very high expression in the Purkinje cells of the cerebellum. InsP3R1 contains a calcium channel in its C-terminal extremity. It has a large N-terminal cytoplasmic region that has the ligand-binding site in the N-terminus and modulatory sites in the middle portion immediately upstream of the channel region. InsP3R1 serves as an intracellular channel that mediates calcium release from the endoplasmic reticulum following stimulation by inositol 1,4,5-trisphosphate.It is involved in the regulation of epithelial secretion of electrolytes and fluid through the interaction with AHCYL1. It also plays a role in ER stress-induced apoptosis wherein cytoplasmic calcium released from the ER triggers apoptosis by the activation of CaM kinase II, eventually leading to the activation of downstream apoptosis pathways. InsP3R1 is phosphorylated by cAMP-dependent protein kinase (PKA), which increases the interaction with inositol 1,4,5-trisphosphate and reduces the interaction with AHCYL1. This prevents the ligand-induced opening of the calcium channels. Calcium appears to inhibit ligand binding to the receptor, most probably by interacting with a distinct calcium-binding protein which then inhibits the receptor. Reduced expression of InsP3R1 has been reported in cisplatin-resistant cancer cells and overexpression of InsP3R1 in resistant cells induces apoptosis and increases sensitivity to cisplatin. (Ref.: Tsunoda T., et al. (2005). Oncogene. 24(8):1396-402; Wang, Y., et al. (2001). Cir. Res. 88(2): 202-209).

Specificity

This polyclonal antibody specifically react with IP3 Receptor 1 in multiple species, but does not recognize IP3R2 and IP3R3. It targets an epitope within 19 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of rat Inositol 1,4,5-trisphosphate receptor type 1 (IP3R1).

Application

Detect Inositol 1,4,5-trisphosphate receptor type 1 using this rabbit polyclonal Anti-IP3R1, Cat. No. ABS2129 that has been tested for use in Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting. ,
Research Category
Signaling
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected IP3R1 in 10 µg of rat and mouse brain microsomal preparation.

Western Blotting Analysis: A representative lot detected IP3R1 in bovine salivary gland, DT40 3KO cells stably expressing IP3R1 (Chandrasekhar, R., et. al. (2016). J Biol Chem. 291(10):4846-60).

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected IP3R1 in human brain and rat brain tissues.

Immunoprecipitation Analysis: A representative lot detected IP3R1 in bovine salivary gland (Chandrasekhar, R., et. al. (2016). J Biol Chem. 291(10):4846-60).

Quality

Evaluated by Western Blotting in rat brain microsomal preparation.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected IP3R1 in 10 µg of rat brain microsomal preparation.

Target description

~300 kDa observed; 313.26 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS with 1% BSA and 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yohannes A Mebratu et al.
Nature communications, 8(1), 803-803 (2017-10-08)
Bik reduces hyperplastic epithelial cells by releasing calcium from endoplasmic reticulum stores and causing apoptosis, but the detailed mechanisms are not known. Here we report that Bik dissociates the Bak/Bcl-2 complex to enrich for ER-associated Bak and interacts with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service