Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

ABN896

Sigma-Aldrich

Anti-LDHA

from rabbit, purified by affinity chromatography

Synonym(s):

L-lactate dehydrogenase A chain, Cell proliferation-inducing gene 19 protein, LDH muscle subunit, LDH-A, LDH-M, Renal carcinoma antigen NY-REN-59

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LDHA(3939)

General description

L-lactate dehydrogenase A chain (EC 1.1.1.27; UniProt P00338; also known as Cell proliferation-inducing gene 19 protein, LDH muscle subunit, LDH-A, LDH-M, Renal carcinoma antigen NY-REN-59) is encoded by the LDHA (also known as GSD11, PIG19) gene (Gene ID 3939) in human. Originally described by Otto Warburg and now commonly known as the Warburg effect refers to the phenomenon that many tumor cells use aerobic glycolysis instead of mitochondrial respiration for energy production to support cell growth and proliferation. Enzymes involved in the complex network of metabolic pathways are therefore being studied as potential targets for cancer treatment. Lactate dehydrogenase (LDH) is a tetrameric enzyme composed of two different subunits, LDHA and LDHB, which can assemble into five different combinations (LDH1 is comprised of four LDHB subunits; LDH2 contains three LDHB and one LDHA subunit; LDH3 contains two LDHB and two LDHA subunits; LDH4 contains one LDHB and three LDHA subunits; LDH5 contains four LDHA subunits). LDHB is ubiquitously expressed and the predominant isoform in heart muscle, while LDHA is the predominant isoform in skeletal muscle and other highly glycolytic tissues. LDH catalyzes the reversible conversion of pyruvate into lactate. An upregulation of LDH therefore reduces the oxygen dependency and ensures an efficient anaerobic/glycolytic metabolism in tumor cells. Upregulated LDHA has been observed in hepatocellular carcinoma, breast, pancreatic, and prostate cancers. Inhibition of LDHA reduced cell malignant transformation and tumor formation. Serum LDHB level is also reported to elevate in lung cancer patients and progressively increase with the clinical stage of non-small cell lung cancer (NSCLC).

Specificity

This rabbit polyclonal targeted epitope region is present in human LDHA spliced isoforms 1, 3, and 4, but absent from isoforms 2 and 5, reported by UniProt (P00338).

Immunogen

KLH-conjugated linear peptide corresponding to a C-terminal region sequence of human LDHA.

Application

Anti-LDHA, Cat. No. ABN896, is a highly specific rabbit polyclonal antibody, that targets L-lactate dehydrogenase A chain and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected LDHA in HUVECs and HeLa cells.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected LDHA in rat testis, human prostate cancer and kidney tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected LDHA in 10 µg of HepG2 cell lysate.

Target description

~37 kDa observed. 36.56/39.71/30.07 kDa (human isoform 1/3/4) and 36.32 kDa (rat) calculated (Met1 removed). Uncharacterized band(s) may appear in some lysates.

Linkage

Replaces: ABN311

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service