Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE68

Sigma-Aldrich

Anti-FoxP1 Antibody

serum, from rabbit

Synonym(s):

forkhead box P1, fork head-related protein like B, glutamine-rich factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXP1(27086)

General description

Forkhead Box P1 (FoxP1) is a member of the FOX family of transcription factors. This protein is known to forms homodimers and heterodimers with family members FoxP2 and FoxP4. The forkhead box family has a broad range of functions. These proteins play important roles in immune response, organ development and the development of cancer. The FoxP1 protein is widely expressed and contains both forkhead (winged-helix) and C2H2 zinc finger nucleic acid-binding motifs. At least four different isoforms that result from alternative splicing of the mRNA transcript have been reported.

Specificity

This antibody recognizes FoxP1.

Immunogen

Epitope: Unknown
GST-tagged recombinant protein corresponding to mouse FoxP1.

Application

Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected FoxP1 in HeLa and NIH/3T3 cells.

Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in IH. (Lu, M.M., et al. (2002). Mechanisms of Development 119S:S197–S202.)
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Developmental Signaling
Use Anti-FoxP1 Antibody (Rabbit Polyclonal Antibody) validated in ICC, WB, IHC to detect FoxP1 also known as forkhead box P1, fork head-related protein like B, glutamine-rich factor 1.

Quality

Evaluated by Western Blot in RAW264.7 cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected FoxP1 on 10 µg of RAW264.7 cell lysate.

Target description

79 kDa observed. Alternative splicing can result in cell type dependent isoforms.

Physical form

Unpurified
Unpurified rabbit polyclonal serum with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
RAW264.7 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shifeng Ling et al.
Stem cell research & therapy, 14(1), 188-188 (2023-07-29)
Mesenchymal stem cells (MSCs) are widely used in a variety of tissue regeneration and clinical trials due to their multiple differentiation potency. However, it remains challenging to maintain their replicative capability during in vitro passaging while preventing their premature cellular
Wei Zhang et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 36(10), 2017-2026 (2021-06-17)
Adiponectin (AdipoQ), a hormone abundantly secreted by adipose tissues, has multiple beneficial functions, including insulin sensitization as well as lipid and glucose metabolism. It has been reported that bone controls energy metabolism through an endocrine-based mechanism. In this study, we
Catherine Konopacki et al.
Nature immunology, 20(2), 232-242 (2019-01-16)
Regulatory T cells (Treg cells), whose differentiation and function are controlled by transcription factor Foxp3, express the closely related family member Foxp1. Here we explored Foxp1 function in Treg cells. We found that a large number of Foxp3-bound genomic sites
Jian He et al.
Cell research, 31(7), 742-757 (2021-01-22)
Human skeletal stem cells (SSCs) have been discovered in fetal and adult long bones. However, the spatiotemporal ontogeny of human embryonic SSCs during early skeletogenesis remains elusive. Here we map the transcriptional landscape of human limb buds and embryonic long
Hanjun Li et al.
The Journal of clinical investigation, 127(4), 1241-1253 (2017-02-28)
A hallmark of aged mesenchymal stem/progenitor cells (MSCs) in bone marrow is the pivot of differentiation potency from osteoblast to adipocyte coupled with a decrease in self-renewal capacity. However, how these cellular events are orchestrated in the aging progress is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service