Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB3094

Sigma-Aldrich

Anti-Orexin-2 Receptor Antibody

Chemicon®, from rabbit

Synonym(s):

OX2R, Hypocretin Receptor 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... HCRTR2(3062)

Specificity

Rat/Human Orexin-2. Several peptides associated with feeding behavior have been reported recently. Orexins (Orexin-A and Orexin-B) are a family of hypothalamic neuropeptides selectively expressed in the hypothalamus (Sakurai et al. 1998). Orexin-A and Orexin-B are derived from the same precursor (Prepro-orexin) by proteolytic cleavage. Prepro-orexin is an 130 aa peptide with a putative 33 aa secretory sequence, a hydrophobic core followed by residues with small polar side chains. The expression was detected in brain and to a small extent in testis (Sakurai et al. 1998). These neuropeptides bind and activate two closely related Orexin receptors--G protein coupled receptors (GPCRs) OX1R and OX2R.

Immunogen

A 19 aa peptide near the C-terminus (cytoplasmic domain) of rat OX2R. The immunogen peptide (Catalog Number AG794) is available to perform preabsorbtion controls.

Application

Detect Orexin-2 Receptor using this Anti-Orexin-2 Receptor Antibody validated for use in ELISA, IH & WB.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Circadian Rhythm & Sleep
Western blot: 1-10 μg/mL using Chemiluminescence technique

ELISA: 1:10,000 - 100,000 against the control peptide (Catalog Number AG794).

Optimal working dilutions must be determined by end user.

Physical form

Affinity Purified 1mg/mL solution in PBS, pH 7.4 and 0.1% BSA

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caterina Squillacioti et al.
Frontiers in veterinary science, 9, 880022-880022 (2022-07-30)
Orexins A (OXA) and B (OXB) and the receptors 1 (OX1R) and 2 (OX2R) for orexins are hypothalamic peptides found in several mammalian organs and participated to the control of a wide assortment of physiological and pathological functions. The distribution
Krister Melén et al.
Journal of translational autoimmunity, 3, 100055-100055 (2020-08-04)
Narcolepsy type 1, likely an immune-mediated disease, is characterized by excessive daytime sleepiness and cataplexy. The disease is strongly associated with human leukocyte antigen (HLA) DQB1∗06:02. A significant increase in the incidence of childhood and adolescent narcolepsy was observed after
P Dehan et al.
The Journal of clinical endocrinology and metabolism, 98(4), 1549-1557 (2013-03-14)
Orexins A and B are neuropeptides that bind and activate 2 types of receptors. In addition to direct action in the brain, the orexinergic system has broader implications in peripheral organs, and it has been proposed to have a role
Chung-Wei Hsu et al.
Brain sciences, 11(12) (2021-12-25)
Orexin-A (OX-A) and orexin-B (OX-B) are neuropeptides produced in the hypothalamus. Preclinical and clinical studies suggest that depression and anxiety are associated with the orexin system. In the current study, we used the learned helplessness (LH) animal model of depression
Qin-Qin Wang et al.
Cellular signalling, 54, 46-58 (2018-11-28)
As G-protein-coupled receptors (GPCRs), 5-hydroxytryptamine 1A receptor (5-HT1AR) and orexin receptor 2 (OX2R) regulate the levels of the cellular downstream molecules. The heterodimers of different GPCRs play important roles in various of neurological diseases. Moreover, 5-HT1AR and OX2R are involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service